Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227523758-227523985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGFG1
Entrez Gene ID
3267
Gene Name
ArfGAP with FG repeats 1
Gene Aliases
HRB, RAB, RIP
UniGene
Hs.352962
Gene Chromosome Location
Chr.2: 227472169-227561222 on Build GRCh38
Transcripts
NM_001135187.1 NM_001135188.1 NM_001135189.1 NM_004504.4 XM_005246516.3 XM_006712478.3 XM_006712479.3 XM_006712480.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6596227 c.421G>A p.A141T Chr.2: 227523806-227523806 on Build GRCh38
1405982 c.443C>T p.A148V Chr.2: 227523828-227523828 on Build GRCh38
3047418 c.406G>A p.V136M Chr.2: 227523791-227523791 on Build GRCh38
4987840 c.409G>A p.A137T Chr.2: 227523794-227523794 on Build GRCh38
3783403 c.526G>A p.G176S Chr.2: 227523911-227523911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770625658 Chr.2: 227523877-227523877 on Build GRCh38
rs192248440 Chr.2: 227523850-227523850 on Build GRCh38
rs761100669 Chr.2: 227523791-227523791 on Build GRCh38
rs183979520 Chr.2: 227523958-227523958 on Build GRCh38
rs187400935 Chr.2: 227523831-227523831 on Build GRCh38
rs778303580 Chr.2: 227523812-227523812 on Build GRCh38
rs746843836 Chr.2: 227523856-227523856 on Build GRCh38
rs374469881 Chr.2: 227523950-227523950 on Build GRCh38
rs754578194 Chr.2: 227523808-227523808 on Build GRCh38
rs185026138 Chr.2: 227523822-227523822 on Build GRCh38
rs767807753 Chr.2: 227523785-227523785 on Build GRCh38
rs752140735 Chr.2: 227523818-227523818 on Build GRCh38
rs781247257 Chr.2: 227523896-227523896 on Build GRCh38
rs772378018 Chr.2: 227523958-227523961 on Build GRCh38
rs200107887 Chr.2: 227523933-227523933 on Build GRCh38
rs933367043 Chr.2: 227523901-227523901 on Build GRCh38
rs565915999 Chr.2: 227523943-227523943 on Build GRCh38
rs750840866 Chr.2: 227523790-227523790 on Build GRCh38
rs777378633 Chr.2: 227523841-227523841 on Build GRCh38
rs147524470 Chr.2: 227523910-227523910 on Build GRCh38
rs61752216 Chr.2: 227523801-227523801 on Build GRCh38
rs370320326 Chr.2: 227523793-227523793 on Build GRCh38
rs921991135 Chr.2: 227523863-227523863 on Build GRCh38
rs943218821 Chr.2: 227523957-227523957 on Build GRCh38
rs745804768 Chr.2: 227523911-227523911 on Build GRCh38

View Full Product Details