Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89100878-89101105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSF3
Entrez Gene ID
197322
Gene Name
acyl-CoA synthetase family member 3
Gene Aliases
-
UniGene
Hs.461727
Gene Chromosome Location
Chr.16: 89093809-89160556 on Build GRCh38
Transcripts
NM_001127214.3 NM_001243279.2 NM_001284316.1 NM_174917.4 NR_104293.1 XM_005256293.2 XM_011522942.1 XM_011522943.1 XM_017023018.1 XM_017023019.1 XM_017023020.1 XM_017023021.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7234594 c.297C>T p.S99S Chr.16: 89100978-89100978 on Build GRCh38
3765952 c.354T>C p.S118S Chr.16: 89101035-89101035 on Build GRCh38
3765951 c.315T>C p.D105D Chr.16: 89100996-89100996 on Build GRCh38
4129507 c.342G>C p.A114A Chr.16: 89101023-89101023 on Build GRCh38
6195252 c.388C>T p.P130S Chr.16: 89101069-89101069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182147718 Chr.16: 89101038-89101038 on Build GRCh38
rs754782061 Chr.16: 89101037-89101037 on Build GRCh38
rs879113913 Chr.16: 89101024-89101024 on Build GRCh38
rs145969050 Chr.16: 89100994-89100994 on Build GRCh38
rs763197756 Chr.16: 89100906-89100906 on Build GRCh38
rs373476048 Chr.16: 89100932-89100932 on Build GRCh38
rs748128555 Chr.16: 89100998-89100998 on Build GRCh38
rs144411003 Chr.16: 89101072-89101072 on Build GRCh38
rs769957763 Chr.16: 89100958-89100958 on Build GRCh38
rs543359504 Chr.16: 89101050-89101051 on Build GRCh38
rs761267255 Chr.16: 89101021-89101021 on Build GRCh38
rs59213357 Chr.16: 89100978-89100978 on Build GRCh38
rs753248595 Chr.16: 89100970-89100970 on Build GRCh38
rs532903196 Chr.16: 89100949-89100949 on Build GRCh38
rs749371364 Chr.16: 89101047-89101047 on Build GRCh38
rs752534981 Chr.16: 89101084-89101084 on Build GRCh38
rs769716004 Chr.16: 89101001-89101001 on Build GRCh38
rs781244439 Chr.16: 89100997-89100997 on Build GRCh38
rs905316715 Chr.16: 89101074-89101074 on Build GRCh38
rs778403709 Chr.16: 89100934-89100934 on Build GRCh38
rs981914227 Chr.16: 89100974-89100974 on Build GRCh38
rs768265862 Chr.16: 89101008-89101009 on Build GRCh38
rs187962814 Chr.16: 89100966-89100966 on Build GRCh38
rs541662926 Chr.16: 89101065-89101065 on Build GRCh38
rs148895267 Chr.16: 89100950-89100950 on Build GRCh38
rs764592417 Chr.16: 89101022-89101022 on Build GRCh38
rs6500527 Chr.16: 89101023-89101023 on Build GRCh38
rs753339356 Chr.16: 89100920-89100920 on Build GRCh38
rs777054379 Chr.16: 89101066-89101066 on Build GRCh38
rs6500526 Chr.16: 89100987-89100987 on Build GRCh38
rs746883692 Chr.16: 89100948-89100948 on Build GRCh38
rs370968781 Chr.16: 89101080-89101080 on Build GRCh38
rs775569136 Chr.16: 89100940-89100940 on Build GRCh38
rs368192538 Chr.16: 89101071-89101071 on Build GRCh38
rs746787960 Chr.16: 89100995-89100995 on Build GRCh38
rs779919537 Chr.16: 89100942-89100942 on Build GRCh38
rs755491777 Chr.16: 89100917-89100917 on Build GRCh38
rs150050697 Chr.16: 89101068-89101068 on Build GRCh38
rs6500528 Chr.16: 89101035-89101035 on Build GRCh38
rs777642279 Chr.16: 89101044-89101044 on Build GRCh38
rs756085657 Chr.16: 89101042-89101042 on Build GRCh38
rs766802453 Chr.16: 89100908-89100908 on Build GRCh38
rs757905943 Chr.16: 89101029-89101029 on Build GRCh38
rs775845691 Chr.16: 89101059-89101059 on Build GRCh38
rs201113468 Chr.16: 89100951-89100951 on Build GRCh38
rs749994320 Chr.16: 89100993-89100993 on Build GRCh38
rs751929678 Chr.16: 89100964-89100964 on Build GRCh38
rs530503915 Chr.16: 89100961-89100961 on Build GRCh38
rs200703917 Chr.16: 89101039-89101039 on Build GRCh38
rs771723461 Chr.16: 89100940-89100940 on Build GRCh38
rs771182485 Chr.16: 89101006-89101006 on Build GRCh38
rs760759040 Chr.16: 89100986-89100986 on Build GRCh38
rs6500529 Chr.16: 89101050-89101050 on Build GRCh38
rs761018548 Chr.16: 89101060-89101060 on Build GRCh38
rs763021932 Chr.16: 89101005-89101005 on Build GRCh38
rs148768970 Chr.16: 89101073-89101073 on Build GRCh38
rs772372919 Chr.16: 89101054-89101054 on Build GRCh38
rs571472953 Chr.16: 89101007-89101007 on Build GRCh38
rs189821127 Chr.16: 89101009-89101009 on Build GRCh38
rs772073893 Chr.16: 89100927-89100927 on Build GRCh38
rs7193255 Chr.16: 89100996-89100996 on Build GRCh38
rs141517318 Chr.16: 89101008-89101008 on Build GRCh38
rs138156311 Chr.16: 89100919-89100919 on Build GRCh38
rs766109997 Chr.16: 89101034-89101034 on Build GRCh38
rs750707295 Chr.16: 89100902-89100902 on Build GRCh38
rs901932797 Chr.16: 89101002-89101002 on Build GRCh38
rs994160606 Chr.16: 89100952-89100952 on Build GRCh38
rs766597613 Chr.16: 89100962-89100962 on Build GRCh38
rs746501634 Chr.16: 89100911-89100911 on Build GRCh38
rs142006073 Chr.16: 89101049-89101050 on Build GRCh38
rs560101290 Chr.16: 89100939-89100939 on Build GRCh38
rs143605434 Chr.16: 89100959-89100959 on Build GRCh38

View Full Product Details