Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 72196820-72197053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASCC1
Entrez Gene ID
51008
Gene Name
activating signal cointegrator 1 complex subunit 1
Gene Aliases
ASC1p50, CGI-18, SMABF2, p50
UniGene
Hs.500007
Gene Chromosome Location
Chr.10: 72096032-72217134 on Build GRCh38
Transcripts
NM_001198798.2 NM_001198799.2 NM_001198800.2 NR_045564.1 XM_006717873.3 XM_011539841.2 XM_011539842.2 XM_017016291.1 XM_017016292.1 XM_017016293.1 XM_017016294.1 XM_017016295.1 XM_017016296.1 XM_017016297.1 XM_017016298.1 XM_017016299.1 XM_017016300.1 XM_017016301.1 XM_017016302.1 XM_017016303.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6129944 c.409C>G p.L137V Chr.10: 72196891-72196891 on Build GRCh38
5014925 c.487C>T p.H163Y Chr.10: 72196897-72196897 on Build GRCh38
3666780 c.378T>C p.T126T Chr.10: 72196922-72196922 on Build GRCh38
3440329 c.439C>T p.Q147* Chr.10: 72196861-72196861 on Build GRCh38
3440326 c.446G>A p.G149E Chr.10: 72196854-72196854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183415577 Chr.10: 72196918-72196918 on Build GRCh38
rs866050664 Chr.10: 72196972-72196972 on Build GRCh38
rs148373391 Chr.10: 72196863-72196863 on Build GRCh38
rs779932786 Chr.10: 72196950-72196950 on Build GRCh38
rs138030710 Chr.10: 72196904-72196904 on Build GRCh38
rs375562737 Chr.10: 72196944-72196944 on Build GRCh38
rs367554892 Chr.10: 72196960-72196960 on Build GRCh38
rs776119295 Chr.10: 72197009-72197009 on Build GRCh38
rs778442657 Chr.10: 72196919-72196919 on Build GRCh38
rs752377351 Chr.10: 72196964-72196964 on Build GRCh38
rs766652867 Chr.10: 72196876-72196876 on Build GRCh38
rs759002398 Chr.10: 72196870-72196870 on Build GRCh38
rs753330159 Chr.10: 72197027-72197027 on Build GRCh38
rs764385166 Chr.10: 72196895-72196895 on Build GRCh38
rs555612625 Chr.10: 72196971-72196971 on Build GRCh38
rs759835418 Chr.10: 72196882-72196882 on Build GRCh38
rs565723980 Chr.10: 72196869-72196869 on Build GRCh38
rs1032751168 Chr.10: 72197018-72197018 on Build GRCh38
rs146356471 Chr.10: 72196925-72196925 on Build GRCh38
rs1045359481 Chr.10: 72196850-72196850 on Build GRCh38
rs774462067 Chr.10: 72196992-72196992 on Build GRCh38
rs747955808 Chr.10: 72196880-72196880 on Build GRCh38
rs768092093 Chr.10: 72196901-72196901 on Build GRCh38
rs774663258 Chr.10: 72197003-72197004 on Build GRCh38
rs573910884 Chr.10: 72196988-72196988 on Build GRCh38
rs937013345 Chr.10: 72196861-72196861 on Build GRCh38
rs1003738261 Chr.10: 72196993-72196993 on Build GRCh38
rs193204024 Chr.10: 72196945-72196945 on Build GRCh38
rs761323613 Chr.10: 72197016-72197016 on Build GRCh38
rs764701971 Chr.10: 72197024-72197024 on Build GRCh38
rs976617998 Chr.10: 72196875-72196875 on Build GRCh38
rs761839119 Chr.10: 72196855-72196855 on Build GRCh38
rs137945482 Chr.10: 72196979-72196979 on Build GRCh38
rs746994660 Chr.10: 72196951-72196951 on Build GRCh38
rs755656113 Chr.10: 72196907-72196907 on Build GRCh38
rs759907999 Chr.10: 72196994-72196994 on Build GRCh38
rs753419523 Chr.10: 72196917-72196917 on Build GRCh38
rs769899560 Chr.10: 72196856-72196856 on Build GRCh38

View Full Product Details