Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68230157-68230420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRP2
Entrez Gene ID
80004
Gene Name
epithelial splicing regulatory protein 2
Gene Aliases
RBM35B
UniGene
Hs.436585
Gene Chromosome Location
Chr.16: 68228547-68236233 on Build GRCh38
Transcripts
NM_024939.2 XM_005256153.4 XM_006721273.3 XM_011523338.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6274604 c.2053C>A p.Q685K Chr.16: 68230400-68230400 on Build GRCh38
6273126 c.2046C>A p.S682R Chr.16: 68230407-68230407 on Build GRCh38
2996294 c.2070C>A p.P690P Chr.16: 68230310-68230310 on Build GRCh38
2996286 c.2137G>T p.E713* Chr.16: 68230243-68230243 on Build GRCh38
6190916 c.2137C>A p.P713T Chr.16: 68230273-68230273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762246969 Chr.16: 68230223-68230223 on Build GRCh38
rs187787483 Chr.16: 68230184-68230184 on Build GRCh38
rs776225879 Chr.16: 68230235-68230235 on Build GRCh38
rs3743737 Chr.16: 68230298-68230298 on Build GRCh38
rs780936867 Chr.16: 68230296-68230296 on Build GRCh38
rs191316190 Chr.16: 68230191-68230191 on Build GRCh38
rs781168292 Chr.16: 68230368-68230368 on Build GRCh38
rs779842960 Chr.16: 68230373-68230373 on Build GRCh38
rs756381379 Chr.16: 68230248-68230248 on Build GRCh38
rs759151563 Chr.16: 68230192-68230192 on Build GRCh38
rs774964049 Chr.16: 68230213-68230213 on Build GRCh38
rs765734681 Chr.16: 68230326-68230327 on Build GRCh38
rs748928247 Chr.16: 68230382-68230382 on Build GRCh38
rs768186292 Chr.16: 68230328-68230328 on Build GRCh38
rs754035265 Chr.16: 68230357-68230357 on Build GRCh38
rs763513287 Chr.16: 68230217-68230217 on Build GRCh38
rs1002896533 Chr.16: 68230249-68230249 on Build GRCh38
rs757298912 Chr.16: 68230258-68230259 on Build GRCh38
rs745541984 Chr.16: 68230273-68230273 on Build GRCh38
rs377274422 Chr.16: 68230252-68230252 on Build GRCh38
rs944062353 Chr.16: 68230397-68230397 on Build GRCh38
rs745806039 Chr.16: 68230369-68230369 on Build GRCh38
rs764247542 Chr.16: 68230198-68230201 on Build GRCh38
rs201109497 Chr.16: 68230360-68230360 on Build GRCh38
rs766855318 Chr.16: 68230352-68230352 on Build GRCh38
rs567277463 Chr.16: 68230381-68230381 on Build GRCh38
rs768230534 Chr.16: 68230394-68230394 on Build GRCh38
rs569178519 Chr.16: 68230267-68230267 on Build GRCh38
rs768479560 Chr.16: 68230340-68230340 on Build GRCh38
rs376551656 Chr.16: 68230339-68230339 on Build GRCh38
rs374441733 Chr.16: 68230187-68230187 on Build GRCh38
rs376539096 Chr.16: 68230183-68230183 on Build GRCh38
rs755179416 Chr.16: 68230359-68230359 on Build GRCh38
rs762348246 Chr.16: 68230326-68230326 on Build GRCh38
rs773762410 Chr.16: 68230321-68230321 on Build GRCh38
rs371553582 Chr.16: 68230266-68230266 on Build GRCh38
rs372754505 Chr.16: 68230372-68230372 on Build GRCh38
rs750857196 Chr.16: 68230243-68230243 on Build GRCh38
rs762587218 Chr.16: 68230216-68230216 on Build GRCh38
rs770807727 Chr.16: 68230275-68230275 on Build GRCh38
rs79902614 Chr.16: 68230310-68230310 on Build GRCh38
rs571268163 Chr.16: 68230188-68230188 on Build GRCh38
rs750931685 Chr.16: 68230334-68230336 on Build GRCh38
rs377038240 Chr.16: 68230206-68230206 on Build GRCh38
rs752044282 Chr.16: 68230218-68230218 on Build GRCh38
rs374092654 Chr.16: 68230251-68230251 on Build GRCh38
rs767951454 Chr.16: 68230225-68230225 on Build GRCh38
rs35355998 Chr.16: 68230309-68230309 on Build GRCh38
rs62637560 Chr.16: 68230306-68230306 on Build GRCh38
rs753925149 Chr.16: 68230229-68230230 on Build GRCh38
rs367569599 Chr.16: 68230361-68230361 on Build GRCh38

View Full Product Details