Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48598061-48598302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB-AS3
Entrez Gene ID
404266
Gene Name
HOXB cluster antisense RNA 3
Gene Aliases
-
UniGene
Hs.660088
Gene Chromosome Location
Chr.17: 48590420-48606414 on Build GRCh38
Transcripts
NR_033201.2 NR_033202.2 NR_033203.1 NR_033204.2 NR_033205.1 NR_110329.1 NR_110330.1 NR_110331.1 NM_018952.4 XM_005257283.3 XM_005257284.3 XM_011524727.2

Gene Symbol
HOXB6
Entrez Gene ID
3216
Gene Name
homeobox B6
Gene Aliases
HOX2, HOX2B, HU-2, Hox-2.2
UniGene
Hs.98428
Gene Chromosome Location
Chr.17: 48595736-48604992 on Build GRCh38
Transcripts
NR_033201.2 NR_033202.2 NR_033203.1 NR_033204.2 NR_033205.1 NR_110329.1 NR_110330.1 NR_110331.1 NM_018952.4 XM_005257283.3 XM_005257284.3 XM_011524727.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6532486 c.9C>G p.S3S Chr.17: 48598142-48598142 on Build GRCh38
6369981 c.55G>T p.E19* Chr.17: 48598096-48598096 on Build GRCh38
4931937 c.7T>A p.S3T Chr.17: 48598144-48598144 on Build GRCh38
5698820 c.70C>T p.Q24* Chr.17: 48598081-48598081 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771378458 Chr.17: 48598174-48598174 on Build GRCh38
rs781003624 Chr.17: 48598162-48598163 on Build GRCh38
rs991824106 Chr.17: 48598140-48598140 on Build GRCh38
rs757108837 Chr.17: 48598115-48598115 on Build GRCh38
rs1011905095 Chr.17: 48598181-48598181 on Build GRCh38
rs538824742 Chr.17: 48598118-48598118 on Build GRCh38
rs765546686 Chr.17: 48598187-48598187 on Build GRCh38
rs965480235 Chr.17: 48598268-48598268 on Build GRCh38
rs751386938 Chr.17: 48598106-48598106 on Build GRCh38
rs1033416823 Chr.17: 48598105-48598105 on Build GRCh38
rs779605276 Chr.17: 48598129-48598129 on Build GRCh38
rs889420397 Chr.17: 48598237-48598237 on Build GRCh38
rs145027311 Chr.17: 48598088-48598088 on Build GRCh38
rs565949558 Chr.17: 48598250-48598250 on Build GRCh38
rs764141745 Chr.17: 48598102-48598102 on Build GRCh38
rs11870680 Chr.17: 48598154-48598154 on Build GRCh38
rs1053349378 Chr.17: 48598164-48598164 on Build GRCh38
rs747431187 Chr.17: 48598167-48598167 on Build GRCh38
rs759873291 Chr.17: 48598186-48598186 on Build GRCh38
rs780798894 Chr.17: 48598117-48598117 on Build GRCh38
rs369696821 Chr.17: 48598104-48598104 on Build GRCh38
rs756777815 Chr.17: 48598212-48598212 on Build GRCh38
rs1006573135 Chr.17: 48598243-48598243 on Build GRCh38
rs748779435 Chr.17: 48598150-48598150 on Build GRCh38
rs1042995995 Chr.17: 48598189-48598189 on Build GRCh38
rs776833019 Chr.17: 48598180-48598180 on Build GRCh38
rs891660129 Chr.17: 48598169-48598169 on Build GRCh38
rs11868402 Chr.17: 48598155-48598155 on Build GRCh38
rs138703831 Chr.17: 48598276-48598276 on Build GRCh38
rs552230381 Chr.17: 48598163-48598163 on Build GRCh38

View Full Product Details