Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40380905-40381150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK4
Entrez Gene ID
147746
Gene Name
homeodomain interacting protein kinase 4
Gene Aliases
-
UniGene
Hs.79363
Gene Chromosome Location
Chr.19: 40379271-40390221 on Build GRCh38
Transcripts
NM_144685.4 XM_006723036.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3534044 c.844G>A p.E282K Chr.19: 40380922-40380922 on Build GRCh38
996661 c.936G>A p.A312A Chr.19: 40381055-40381055 on Build GRCh38
4078194 c.866A>G p.D289G Chr.19: 40381125-40381125 on Build GRCh38
6496001 c.1011C>T p.S337S Chr.19: 40380980-40380980 on Build GRCh38
6557907 c.1056G>A p.L352L Chr.19: 40380935-40380935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029674069 Chr.19: 40380953-40380953 on Build GRCh38
rs1010853279 Chr.19: 40381048-40381048 on Build GRCh38
rs976136541 Chr.19: 40381114-40381114 on Build GRCh38
rs760809424 Chr.19: 40381014-40381014 on Build GRCh38
rs144871802 Chr.19: 40381051-40381051 on Build GRCh38
rs570485639 Chr.19: 40380926-40380926 on Build GRCh38
rs745936286 Chr.19: 40381068-40381068 on Build GRCh38
rs549775320 Chr.19: 40381025-40381025 on Build GRCh38
rs763464903 Chr.19: 40381087-40381087 on Build GRCh38
rs774483576 Chr.19: 40381062-40381062 on Build GRCh38
rs147334390 Chr.19: 40380949-40380949 on Build GRCh38
rs772511144 Chr.19: 40381104-40381104 on Build GRCh38
rs752178632 Chr.19: 40381092-40381092 on Build GRCh38
rs757181826 Chr.19: 40381126-40381126 on Build GRCh38
rs374741709 Chr.19: 40381045-40381045 on Build GRCh38
rs368679795 Chr.19: 40380952-40380952 on Build GRCh38
rs531192572 Chr.19: 40381022-40381022 on Build GRCh38
rs775458172 Chr.19: 40381078-40381078 on Build GRCh38
rs200426473 Chr.19: 40380992-40380992 on Build GRCh38
rs764500728 Chr.19: 40380933-40380933 on Build GRCh38
rs527880457 Chr.19: 40380974-40380974 on Build GRCh38
rs780574061 Chr.19: 40381049-40381049 on Build GRCh38
rs907995369 Chr.19: 40381098-40381098 on Build GRCh38
rs867325766 Chr.19: 40381027-40381027 on Build GRCh38
rs754317067 Chr.19: 40380934-40380934 on Build GRCh38
rs754550193 Chr.19: 40381046-40381046 on Build GRCh38
rs79330270 Chr.19: 40381080-40381080 on Build GRCh38
rs775846215 Chr.19: 40381009-40381009 on Build GRCh38
rs749185979 Chr.19: 40380964-40380964 on Build GRCh38
rs11670988 Chr.19: 40381086-40381086 on Build GRCh38
rs371188356 Chr.19: 40381015-40381015 on Build GRCh38
rs145470994 Chr.19: 40381055-40381055 on Build GRCh38
rs757786495 Chr.19: 40380939-40380939 on Build GRCh38
rs1040483836 Chr.19: 40380968-40380968 on Build GRCh38
rs370183538 Chr.19: 40381121-40381121 on Build GRCh38
rs756557433 Chr.19: 40380929-40380929 on Build GRCh38
rs772895580 Chr.19: 40380963-40380963 on Build GRCh38
rs140926477 Chr.19: 40381102-40381102 on Build GRCh38
rs767834907 Chr.19: 40381003-40381003 on Build GRCh38
rs139337380 Chr.19: 40381069-40381069 on Build GRCh38
rs760914615 Chr.19: 40381079-40381079 on Build GRCh38
rs554119598 Chr.19: 40381061-40381061 on Build GRCh38
rs893696566 Chr.19: 40381035-40381035 on Build GRCh38
rs770966226 Chr.19: 40381056-40381056 on Build GRCh38
rs55869814 Chr.19: 40380959-40380959 on Build GRCh38
rs779503136 Chr.19: 40380944-40380944 on Build GRCh38
rs755743447 Chr.19: 40381052-40381052 on Build GRCh38
rs567036647 Chr.19: 40380999-40380999 on Build GRCh38
rs769566712 Chr.19: 40380960-40380960 on Build GRCh38
rs777338552 Chr.19: 40381097-40381097 on Build GRCh38
rs140411181 Chr.19: 40381107-40381107 on Build GRCh38
rs144600040 Chr.19: 40381095-40381095 on Build GRCh38
rs985974580 Chr.19: 40380977-40380977 on Build GRCh38

View Full Product Details