Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 5771236-5771491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARS2
Entrez Gene ID
10667
Gene Name
phenylalanyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD14, FARS1, HSPC320, PheRS
UniGene
Hs.484547
Gene Chromosome Location
Chr.6: 5261001-5771592 on Build GRCh38
Transcripts
NM_001318872.1 NM_006567.4 XM_005248812.3 XM_017010186.1 XM_017010187.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3159950 c.1243C>T p.R415C Chr.6: 5771316-5771316 on Build GRCh38
1329579 c.1304A>T p.Q435L Chr.6: 5771377-5771377 on Build GRCh38
1329580 c.1226A>G p.K409R Chr.6: 5771299-5771299 on Build GRCh38
3629103 c.1262T>A p.M421K Chr.6: 5771335-5771335 on Build GRCh38
1568294 c.1267C>T p.R423W Chr.6: 5771340-5771340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143662929 Chr.6: 5771346-5771346 on Build GRCh38
rs201607143 Chr.6: 5771393-5771393 on Build GRCh38
rs945267016 Chr.6: 5771417-5771417 on Build GRCh38
rs372744573 Chr.6: 5771460-5771460 on Build GRCh38
rs1037000506 Chr.6: 5771263-5771263 on Build GRCh38
rs764850280 Chr.6: 5771464-5771464 on Build GRCh38
rs535897485 Chr.6: 5771418-5771418 on Build GRCh38
rs774824864 Chr.6: 5771378-5771378 on Build GRCh38
rs772372392 Chr.6: 5771386-5771386 on Build GRCh38
rs770283015 Chr.6: 5771287-5771287 on Build GRCh38
rs776691105 Chr.6: 5771408-5771408 on Build GRCh38
rs965780712 Chr.6: 5771381-5771381 on Build GRCh38
rs763330995 Chr.6: 5771404-5771404 on Build GRCh38
rs751041525 Chr.6: 5771441-5771441 on Build GRCh38
rs372054960 Chr.6: 5771293-5771293 on Build GRCh38
rs750200609 Chr.6: 5771428-5771428 on Build GRCh38
rs761214773 Chr.6: 5771435-5771435 on Build GRCh38
rs373806299 Chr.6: 5771260-5771260 on Build GRCh38
rs763952391 Chr.6: 5771402-5771402 on Build GRCh38
rs759833179 Chr.6: 5771316-5771316 on Build GRCh38
rs775690041 Chr.6: 5771328-5771328 on Build GRCh38
rs753581476 Chr.6: 5771331-5771331 on Build GRCh38
rs975048540 Chr.6: 5771405-5771405 on Build GRCh38
rs148921184 Chr.6: 5771341-5771341 on Build GRCh38
rs748697780 Chr.6: 5771282-5771282 on Build GRCh38
rs778281728 Chr.6: 5771353-5771353 on Build GRCh38
rs144758353 Chr.6: 5771317-5771317 on Build GRCh38
rs145697325 Chr.6: 5771340-5771340 on Build GRCh38
rs142426372 Chr.6: 5771303-5771303 on Build GRCh38
rs189168256 Chr.6: 5771324-5771324 on Build GRCh38
rs757681409 Chr.6: 5771466-5771466 on Build GRCh38
rs746478164 Chr.6: 5771348-5771348 on Build GRCh38
rs148155284 Chr.6: 5771384-5771384 on Build GRCh38
rs559504494 Chr.6: 5771368-5771368 on Build GRCh38
rs895849223 Chr.6: 5771267-5771267 on Build GRCh38
rs200232967 Chr.6: 5771433-5771433 on Build GRCh38
rs1000376824 Chr.6: 5771323-5771323 on Build GRCh38
rs761654230 Chr.6: 5771414-5771414 on Build GRCh38
rs779539585 Chr.6: 5771276-5771276 on Build GRCh38
rs367852797 Chr.6: 5771273-5771273 on Build GRCh38
rs202183509 Chr.6: 5771329-5771329 on Build GRCh38
rs368616229 Chr.6: 5771456-5771456 on Build GRCh38
rs749597978 Chr.6: 5771295-5771295 on Build GRCh38
rs80085972 Chr.6: 5771301-5771301 on Build GRCh38
rs758019336 Chr.6: 5771342-5771342 on Build GRCh38
rs749743875 Chr.6: 5771366-5771366 on Build GRCh38
rs1014916887 Chr.6: 5771286-5771286 on Build GRCh38
rs754409124 Chr.6: 5771463-5771463 on Build GRCh38
rs760743151 Chr.6: 5771401-5771401 on Build GRCh38
rs760473865 Chr.6: 5771259-5771259 on Build GRCh38
rs754572509 Chr.6: 5771443-5771443 on Build GRCh38
rs756580731 Chr.6: 5771350-5771350 on Build GRCh38
rs374009001 Chr.6: 5771268-5771268 on Build GRCh38
rs757022549 Chr.6: 5771334-5771334 on Build GRCh38
rs984460938 Chr.6: 5771294-5771294 on Build GRCh38

View Full Product Details