Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150731013-150731238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCTN4
Entrez Gene ID
51164
Gene Name
dynactin subunit 4
Gene Aliases
DYN4, P62
UniGene
Hs.675564
Gene Chromosome Location
Chr.5: 150708747-150759095 on Build GRCh38
Transcripts
NM_001135643.1 NM_001135644.1 NM_016221.3 XM_005268457.1 XM_011537644.1 XM_011537645.1 XM_011537646.1 XM_011537647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6361735 c.652C>T p.P218S Chr.5: 150731116-150731116 on Build GRCh38
7199124 c.614T>C p.L205P Chr.5: 150731154-150731154 on Build GRCh38
1064441 c.633G>T p.Q211H Chr.5: 150731135-150731135 on Build GRCh38
1157741 c.707C>A p.P236Q Chr.5: 150731061-150731061 on Build GRCh38
4676629 c.699T>C p.Y233Y Chr.5: 150731069-150731069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543351232 Chr.5: 150731070-150731070 on Build GRCh38
rs758253397 Chr.5: 150731106-150731106 on Build GRCh38
rs369098676 Chr.5: 150731062-150731062 on Build GRCh38
rs372504610 Chr.5: 150731115-150731115 on Build GRCh38
rs763366168 Chr.5: 150731060-150731060 on Build GRCh38
rs917692142 Chr.5: 150731079-150731079 on Build GRCh38
rs767813564 Chr.5: 150731177-150731177 on Build GRCh38
rs769574064 Chr.5: 150731132-150731132 on Build GRCh38
rs1048310787 Chr.5: 150731190-150731190 on Build GRCh38
rs764355735 Chr.5: 150731186-150731186 on Build GRCh38
rs759987013 Chr.5: 150731166-150731166 on Build GRCh38
rs150127862 Chr.5: 150731123-150731123 on Build GRCh38
rs747539914 Chr.5: 150731077-150731077 on Build GRCh38
rs774175825 Chr.5: 150731170-150731174 on Build GRCh38
rs780084125 Chr.5: 150731107-150731107 on Build GRCh38
rs754780098 Chr.5: 150731112-150731112 on Build GRCh38
rs766970015 Chr.5: 150731061-150731061 on Build GRCh38
rs750811794 Chr.5: 150731169-150731170 on Build GRCh38
rs762066195 Chr.5: 150731158-150731166 on Build GRCh38
rs887516461 Chr.5: 150731169-150731169 on Build GRCh38
rs764097747 Chr.5: 150731086-150731086 on Build GRCh38
rs775839716 Chr.5: 150731179-150731179 on Build GRCh38
rs187193062 Chr.5: 150731138-150731138 on Build GRCh38
rs752872521 Chr.5: 150731073-150731073 on Build GRCh38
rs548801868 Chr.5: 150731150-150731150 on Build GRCh38
rs754012986 Chr.5: 150731088-150731088 on Build GRCh38
rs948550308 Chr.5: 150731100-150731100 on Build GRCh38
rs145810652 Chr.5: 150731069-150731069 on Build GRCh38
rs756345731 Chr.5: 150731076-150731076 on Build GRCh38
rs748881084 Chr.5: 150731140-150731140 on Build GRCh38
rs376698684 Chr.5: 150731163-150731163 on Build GRCh38
rs778382204 Chr.5: 150731058-150731058 on Build GRCh38
rs757394889 Chr.5: 150731200-150731200 on Build GRCh38
rs200122349 Chr.5: 150731192-150731192 on Build GRCh38
rs770856799 Chr.5: 150731207-150731207 on Build GRCh38
rs915725637 Chr.5: 150731074-150731074 on Build GRCh38
rs200735104 Chr.5: 150731089-150731089 on Build GRCh38
rs746746438 Chr.5: 150731111-150731111 on Build GRCh38
rs781156279 Chr.5: 150731121-150731121 on Build GRCh38
rs754977483 Chr.5: 150731208-150731208 on Build GRCh38
rs765262876 Chr.5: 150731201-150731201 on Build GRCh38
rs201517746 Chr.5: 150731151-150731151 on Build GRCh38

View Full Product Details