Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 94558088-94558333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HELLS
Entrez Gene ID
3070
Gene Name
helicase, lymphoid-specific
Gene Aliases
ICF4, LSH, Nbla10143, PASG, SMARCA6
UniGene
Hs.463677
Gene Chromosome Location
Chr.10: 94545767-94613905 on Build GRCh38
Transcripts
NM_001289067.1 NM_001289068.1 NM_001289069.1 NM_001289070.1 NM_001289071.1 NM_001289072.1 NM_001289073.1 NM_001289074.1 NM_001289075.1 NM_018063.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4759057 c.284A>C p.K95T Chr.10: 94558146-94558146 on Build GRCh38
6326652 c.302A>G p.E101G Chr.10: 94558164-94558164 on Build GRCh38
7124670 c.309A>T p.K103N Chr.10: 94558171-94558171 on Build GRCh38
3791047 c.309A>T p.K103N Chr.10: 94558171-94558171 on Build GRCh38
6401677 c.289A>G p.K97E Chr.10: 94558151-94558151 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751905411 Chr.10: 94558153-94558153 on Build GRCh38
rs541785450 Chr.10: 94558143-94558143 on Build GRCh38
rs773663434 Chr.10: 94558121-94558121 on Build GRCh38
rs368577001 Chr.10: 94558210-94558210 on Build GRCh38
rs375581821 Chr.10: 94558208-94558208 on Build GRCh38
rs761369016 Chr.10: 94558228-94558228 on Build GRCh38
rs757275996 Chr.10: 94558199-94558199 on Build GRCh38
rs753013064 Chr.10: 94558176-94558176 on Build GRCh38
rs767602146 Chr.10: 94558175-94558175 on Build GRCh38
rs373528769 Chr.10: 94558119-94558119 on Build GRCh38
rs766864753 Chr.10: 94558238-94558238 on Build GRCh38
rs942532066 Chr.10: 94558265-94558265 on Build GRCh38
rs562051491 Chr.10: 94558211-94558211 on Build GRCh38
rs571640753 Chr.10: 94558168-94558168 on Build GRCh38
rs945802648 Chr.10: 94558165-94558165 on Build GRCh38
rs780629297 Chr.10: 94558200-94558200 on Build GRCh38
rs192831570 Chr.10: 94558213-94558213 on Build GRCh38
rs755435962 Chr.10: 94558160-94558160 on Build GRCh38
rs1042263693 Chr.10: 94558278-94558278 on Build GRCh38
rs377378282 Chr.10: 94558179-94558179 on Build GRCh38
rs370163100 Chr.10: 94558215-94558215 on Build GRCh38
rs756098021 Chr.10: 94558177-94558177 on Build GRCh38
rs375515515 Chr.10: 94558217-94558217 on Build GRCh38
rs778030963 Chr.10: 94558178-94558178 on Build GRCh38
rs201455336 Chr.10: 94558141-94558141 on Build GRCh38
rs551635219 Chr.10: 94558117-94558117 on Build GRCh38
rs1039553786 Chr.10: 94558281-94558286 on Build GRCh38
rs370761610 Chr.10: 94558205-94558205 on Build GRCh38

View Full Product Details