Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28532230-28532503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPX5
Entrez Gene ID
2880
Gene Name
glutathione peroxidase 5
Gene Aliases
HEL-S-75p
UniGene
Hs.248129
Gene Chromosome Location
Chr.6: 28526012-28534951 on Build GRCh38
Transcripts
NM_001509.2 NM_003996.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4389686 c.406G>T p.E136* Chr.6: 28532367-28532367 on Build GRCh38
5737514 c.436C>G p.Q146E Chr.6: 28532397-28532397 on Build GRCh38
1077140 c.440A>C p.K147T Chr.6: 28532401-28532401 on Build GRCh38
335920 c.438G>C p.Q146H Chr.6: 28532399-28532399 on Build GRCh38
6479113 c.395T>C p.F132S Chr.6: 28532356-28532356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750072388 Chr.6: 28532339-28532339 on Build GRCh38
rs756781549 Chr.6: 28532322-28532322 on Build GRCh38
rs985716726 Chr.6: 28532385-28532385 on Build GRCh38
rs377207549 Chr.6: 28532373-28532373 on Build GRCh38
rs750719954 Chr.6: 28532482-28532482 on Build GRCh38
rs755785017 Chr.6: 28532459-28532459 on Build GRCh38
rs750977318 Chr.6: 28532314-28532314 on Build GRCh38
rs151120438 Chr.6: 28532425-28532425 on Build GRCh38
rs773318023 Chr.6: 28532409-28532409 on Build GRCh38
rs368191059 Chr.6: 28532307-28532307 on Build GRCh38
rs762750489 Chr.6: 28532427-28532427 on Build GRCh38
rs138279209 Chr.6: 28532329-28532329 on Build GRCh38
rs930043491 Chr.6: 28532451-28532451 on Build GRCh38
rs955761714 Chr.6: 28532365-28532365 on Build GRCh38
rs764098248 Chr.6: 28532312-28532312 on Build GRCh38
rs760952126 Chr.6: 28532282-28532282 on Build GRCh38
rs747209170 Chr.6: 28532410-28532410 on Build GRCh38
rs949269120 Chr.6: 28532483-28532483 on Build GRCh38
rs931047979 Chr.6: 28532477-28532477 on Build GRCh38
rs778478093 Chr.6: 28532272-28532274 on Build GRCh38
rs987068611 Chr.6: 28532374-28532374 on Build GRCh38
rs368538290 Chr.6: 28532447-28532447 on Build GRCh38
rs748659739 Chr.6: 28532389-28532389 on Build GRCh38
rs942664050 Chr.6: 28532379-28532379 on Build GRCh38
rs769189 Chr.6: 28532458-28532458 on Build GRCh38
rs754590413 Chr.6: 28532272-28532272 on Build GRCh38
rs779483762 Chr.6: 28532381-28532381 on Build GRCh38
rs771175181 Chr.6: 28532415-28532415 on Build GRCh38
rs911500268 Chr.6: 28532377-28532377 on Build GRCh38
rs571923326 Chr.6: 28532449-28532449 on Build GRCh38
rs775287683 Chr.6: 28532302-28532302 on Build GRCh38
rs761375946 Chr.6: 28532395-28532395 on Build GRCh38
rs767033767 Chr.6: 28532452-28532452 on Build GRCh38
rs772188006 Chr.6: 28532394-28532394 on Build GRCh38
rs375980523 Chr.6: 28532290-28532290 on Build GRCh38
rs557024169 Chr.6: 28532481-28532481 on Build GRCh38
rs376005683 Chr.6: 28532446-28532446 on Build GRCh38
rs375591565 Chr.6: 28532298-28532298 on Build GRCh38
rs200652573 Chr.6: 28532366-28532366 on Build GRCh38
rs777068203 Chr.6: 28532258-28532258 on Build GRCh38
rs747541520 Chr.6: 28532448-28532449 on Build GRCh38
rs1048162970 Chr.6: 28532478-28532480 on Build GRCh38
rs565977318 Chr.6: 28532378-28532378 on Build GRCh38
rs755883501 Chr.6: 28532342-28532342 on Build GRCh38
rs780514308 Chr.6: 28532359-28532359 on Build GRCh38
rs766130384 Chr.6: 28532466-28532466 on Build GRCh38
rs754193658 Chr.6: 28532472-28532472 on Build GRCh38

View Full Product Details