Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57155194-57155439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUXA
Entrez Gene ID
503835
Gene Name
double homeobox A
Gene Aliases
-
UniGene
Hs.585857
Gene Chromosome Location
Chr.19: 57151726-57167488 on Build GRCh38
Transcripts
NM_001012729.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1712980 c.529C>T p.P177S Chr.19: 57155282-57155282 on Build GRCh38
4081982 c.498G>A p.A166A Chr.19: 57155313-57155313 on Build GRCh38
1526750 c.455G>A p.R152Q Chr.19: 57155356-57155356 on Build GRCh38
6414138 c.483A>T p.K161N Chr.19: 57155328-57155328 on Build GRCh38
6650425 c.536G>C p.G179A Chr.19: 57155275-57155275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377293882 Chr.19: 57155237-57155237 on Build GRCh38
rs753681514 Chr.19: 57155310-57155310 on Build GRCh38
rs774429337 Chr.19: 57155327-57155327 on Build GRCh38
rs201120927 Chr.19: 57155245-57155245 on Build GRCh38
rs369866205 Chr.19: 57155230-57155230 on Build GRCh38
rs752626275 Chr.19: 57155275-57155275 on Build GRCh38
rs34358992 Chr.19: 57155221-57155222 on Build GRCh38
rs114533845 Chr.19: 57155390-57155390 on Build GRCh38
rs897566009 Chr.19: 57155408-57155408 on Build GRCh38
rs376483102 Chr.19: 57155276-57155276 on Build GRCh38
rs374188662 Chr.19: 57155231-57155231 on Build GRCh38
rs768661356 Chr.19: 57155253-57155253 on Build GRCh38
rs775922541 Chr.19: 57155249-57155249 on Build GRCh38
rs781705411 Chr.19: 57155241-57155241 on Build GRCh38
rs749793051 Chr.19: 57155321-57155321 on Build GRCh38
rs564097384 Chr.19: 57155260-57155260 on Build GRCh38
rs66590417 Chr.19: 57155223-57155224 on Build GRCh38
rs767670445 Chr.19: 57155262-57155262 on Build GRCh38
rs938352420 Chr.19: 57155398-57155398 on Build GRCh38
rs778649278 Chr.19: 57155229-57155229 on Build GRCh38
rs866701297 Chr.19: 57155282-57155282 on Build GRCh38
rs772135803 Chr.19: 57155356-57155356 on Build GRCh38
rs761814715 Chr.19: 57155337-57155337 on Build GRCh38
rs780439658 Chr.19: 57155314-57155314 on Build GRCh38
rs186207313 Chr.19: 57155217-57155217 on Build GRCh38
rs765067191 Chr.19: 57155267-57155267 on Build GRCh38
rs368356011 Chr.19: 57155407-57155407 on Build GRCh38
rs764978802 Chr.19: 57155399-57155399 on Build GRCh38
rs965395752 Chr.19: 57155261-57155261 on Build GRCh38
rs780921059 Chr.19: 57155303-57155303 on Build GRCh38
rs865931856 Chr.19: 57155336-57155336 on Build GRCh38
rs763498462 Chr.19: 57155251-57155251 on Build GRCh38
rs751062136 Chr.19: 57155281-57155281 on Build GRCh38
rs529157053 Chr.19: 57155232-57155232 on Build GRCh38
rs369304573 Chr.19: 57155242-57155242 on Build GRCh38
rs370297842 Chr.19: 57155313-57155313 on Build GRCh38
rs916849995 Chr.19: 57155338-57155338 on Build GRCh38
rs756981949 Chr.19: 57155299-57155299 on Build GRCh38
rs532749484 Chr.19: 57155266-57155266 on Build GRCh38
rs759324672 Chr.19: 57155339-57155342 on Build GRCh38
rs1050328957 Chr.19: 57155334-57155334 on Build GRCh38
rs958200676 Chr.19: 57155311-57155311 on Build GRCh38
rs1020058981 Chr.19: 57155252-57155252 on Build GRCh38
rs753635978 Chr.19: 57155265-57155265 on Build GRCh38
rs747677797 Chr.19: 57155258-57155258 on Build GRCh38
rs745507201 Chr.19: 57155310-57155310 on Build GRCh38
rs766150468 Chr.19: 57155296-57155298 on Build GRCh38
rs765251182 Chr.19: 57155408-57155409 on Build GRCh38
rs773479524 Chr.19: 57155357-57155357 on Build GRCh38
rs144291179 Chr.19: 57155279-57155279 on Build GRCh38

View Full Product Details