Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123097732-123098005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLMP
Entrez Gene ID
79827
Gene Name
CXADR like membrane protein
Gene Aliases
ACAM, ASAM, CSBM, CSBS
UniGene
Hs.591949
Gene Chromosome Location
Chr.11: 123072006-123195305 on Build GRCh38
Transcripts
NM_024769.3 XM_011542998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6630540 c.162C>G p.T54T Chr.11: 123097819-123097819 on Build GRCh38
3444501 c.51G>A p.L17L Chr.11: 123097930-123097930 on Build GRCh38
4518885 c.84_85GG>AA p.E29K Chr.11: 123097896-123097897 on Build GRCh38
3444500 c.174G>A p.G58G Chr.11: 123097807-123097807 on Build GRCh38
6297938 c.137C>A p.T46N Chr.11: 123097844-123097844 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768030215 Chr.11: 123097794-123097794 on Build GRCh38
rs778669110 Chr.11: 123097767-123097767 on Build GRCh38
rs774689924 Chr.11: 123097815-123097815 on Build GRCh38
rs749716633 Chr.11: 123097891-123097891 on Build GRCh38
rs1015313410 Chr.11: 123097783-123097783 on Build GRCh38
rs759661592 Chr.11: 123097933-123097933 on Build GRCh38
rs770277098 Chr.11: 123097800-123097800 on Build GRCh38
rs751141439 Chr.11: 123097861-123097861 on Build GRCh38
rs770453109 Chr.11: 123097887-123097887 on Build GRCh38
rs775441849 Chr.11: 123097975-123097975 on Build GRCh38
rs779732527 Chr.11: 123097859-123097859 on Build GRCh38
rs533120129 Chr.11: 123097764-123097764 on Build GRCh38
rs987156679 Chr.11: 123097854-123097854 on Build GRCh38
rs764138366 Chr.11: 123097761-123097761 on Build GRCh38
rs771389106 Chr.11: 123097897-123097897 on Build GRCh38
rs760179693 Chr.11: 123097891-123097892 on Build GRCh38
rs140007158 Chr.11: 123097890-123097890 on Build GRCh38
rs879253855 Chr.11: 123097954-123097954 on Build GRCh38
rs367573793 Chr.11: 123097789-123097789 on Build GRCh38
rs745608024 Chr.11: 123097769-123097769 on Build GRCh38
rs750253672 Chr.11: 123097848-123097848 on Build GRCh38
rs146055456 Chr.11: 123097818-123097818 on Build GRCh38
rs200965881 Chr.11: 123097967-123097967 on Build GRCh38
rs754960652 Chr.11: 123097755-123097755 on Build GRCh38
rs774860551 Chr.11: 123097761-123097762 on Build GRCh38
rs796648886 Chr.11: 123097907-123097907 on Build GRCh38
rs767487653 Chr.11: 123097819-123097819 on Build GRCh38
rs765094907 Chr.11: 123097845-123097845 on Build GRCh38
rs17850493 Chr.11: 123097962-123097962 on Build GRCh38
rs748588218 Chr.11: 123097797-123097797 on Build GRCh38
rs773714811 Chr.11: 123097801-123097801 on Build GRCh38
rs17856305 Chr.11: 123097956-123097956 on Build GRCh38
rs987761261 Chr.11: 123097834-123097834 on Build GRCh38
rs199765062 Chr.11: 123097902-123097902 on Build GRCh38
rs113227209 Chr.11: 123097788-123097788 on Build GRCh38
rs747626474 Chr.11: 123097882-123097882 on Build GRCh38
rs753682749 Chr.11: 123097763-123097763 on Build GRCh38
rs780580116 Chr.11: 123097872-123097872 on Build GRCh38
rs868240183 Chr.11: 123097890-123097891 on Build GRCh38
rs753874910 Chr.11: 123097820-123097820 on Build GRCh38
rs763368614 Chr.11: 123097813-123097813 on Build GRCh38
rs758053993 Chr.11: 123097773-123097773 on Build GRCh38
rs781743198 Chr.11: 123097796-123097796 on Build GRCh38
rs748245441 Chr.11: 123097822-123097822 on Build GRCh38

View Full Product Details