Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 80393038-80393295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA2
Entrez Gene ID
1496
Gene Name
catenin alpha 2
Gene Aliases
CAP-R, CAPR, CT114, CTNR
UniGene
Hs.167368
Gene Chromosome Location
Chr.2: 79512934-80648868 on Build GRCh38
Transcripts
NM_001164883.1 NM_001282597.2 NM_001282598.1 NM_001282599.1 NM_001282600.1 NM_001320810.1 NM_004389.3 XM_011532555.2 XM_011532556.2 XM_017003403.1 XM_017003404.1 XM_017003405.1 XM_017003406.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
122409 c.1104G>C p.K368N Chr.2: 80393258-80393258 on Build GRCh38
4141263 c.1107G>A p.M369I Chr.2: 80393261-80393261 on Build GRCh38
4943311 c.1214A>G p.K405R Chr.2: 80393266-80393266 on Build GRCh38
3583596 c.1171G>A p.E391K Chr.2: 80393223-80393223 on Build GRCh38
6467430 c.1110T>A p.T370T Chr.2: 80393264-80393264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770693896 Chr.2: 80393244-80393244 on Build GRCh38
rs373365115 Chr.2: 80393230-80393230 on Build GRCh38
rs3770306 Chr.2: 80393110-80393110 on Build GRCh38
rs1055276843 Chr.2: 80393148-80393148 on Build GRCh38
rs755532184 Chr.2: 80393189-80393189 on Build GRCh38
rs771600832 Chr.2: 80393206-80393206 on Build GRCh38
rs756459331 Chr.2: 80393165-80393165 on Build GRCh38
rs61754542 Chr.2: 80393245-80393245 on Build GRCh38
rs140755127 Chr.2: 80393147-80393147 on Build GRCh38
rs17019148 Chr.2: 80393082-80393082 on Build GRCh38
rs1004697998 Chr.2: 80393139-80393139 on Build GRCh38
rs1056033271 Chr.2: 80393078-80393078 on Build GRCh38
rs1014222187 Chr.2: 80393083-80393083 on Build GRCh38
rs774544806 Chr.2: 80393264-80393264 on Build GRCh38
rs765337293 Chr.2: 80393253-80393253 on Build GRCh38
rs772870147 Chr.2: 80393224-80393224 on Build GRCh38
rs112051787 Chr.2: 80393129-80393129 on Build GRCh38
rs553810410 Chr.2: 80393126-80393126 on Build GRCh38
rs747695845 Chr.2: 80393194-80393194 on Build GRCh38
rs940045540 Chr.2: 80393084-80393084 on Build GRCh38
rs762280558 Chr.2: 80393140-80393141 on Build GRCh38
rs1032651242 Chr.2: 80393218-80393218 on Build GRCh38
rs369970894 Chr.2: 80393248-80393248 on Build GRCh38
rs565585755 Chr.2: 80393249-80393249 on Build GRCh38
rs13390195 Chr.2: 80393159-80393159 on Build GRCh38
rs780399155 Chr.2: 80393172-80393172 on Build GRCh38
rs201609332 Chr.2: 80393175-80393175 on Build GRCh38
rs998503394 Chr.2: 80393204-80393204 on Build GRCh38
rs750734155 Chr.2: 80393164-80393164 on Build GRCh38
rs778390882 Chr.2: 80393191-80393191 on Build GRCh38

View Full Product Details