Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155287108-155287375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCN3
Entrez Gene ID
57657
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
Gene Aliases
-
UniGene
Hs.706960
Gene Chromosome Location
Chr.1: 155277427-155289848 on Build GRCh38
Transcripts
NM_020897.2 NR_073074.1 XM_011509816.2 XM_011509817.2 XM_011509818.2 XM_017001918.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
896867 c.1631T>C p.L544P Chr.1: 155287326-155287326 on Build GRCh38
3474924 c.1585T>C p.F529L Chr.1: 155287280-155287280 on Build GRCh38
3474925 c.1636C>T p.R546C Chr.1: 155287331-155287331 on Build GRCh38
7200977 c.1641C>G p.I547M Chr.1: 155287336-155287336 on Build GRCh38
1688719 c.1579G>A p.E527K Chr.1: 155287274-155287274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373228846 Chr.1: 155287334-155287334 on Build GRCh38
rs112049548 Chr.1: 155287215-155287215 on Build GRCh38
rs199859586 Chr.1: 155287288-155287288 on Build GRCh38
rs931531090 Chr.1: 155287197-155287197 on Build GRCh38
rs201531495 Chr.1: 155287293-155287293 on Build GRCh38
rs773423110 Chr.1: 155287342-155287342 on Build GRCh38
rs144688517 Chr.1: 155287322-155287322 on Build GRCh38
rs946623627 Chr.1: 155287232-155287232 on Build GRCh38
rs752827015 Chr.1: 155287141-155287141 on Build GRCh38
rs200418562 Chr.1: 155287353-155287353 on Build GRCh38
rs776866836 Chr.1: 155287336-155287336 on Build GRCh38
rs141800759 Chr.1: 155287248-155287248 on Build GRCh38
rs758961759 Chr.1: 155287208-155287208 on Build GRCh38
rs770967873 Chr.1: 155287251-155287251 on Build GRCh38
rs765497777 Chr.1: 155287233-155287233 on Build GRCh38
rs762732730 Chr.1: 155287249-155287249 on Build GRCh38
rs61742422 Chr.1: 155287303-155287303 on Build GRCh38
rs868827483 Chr.1: 155287252-155287252 on Build GRCh38
rs12402606 Chr.1: 155287191-155287191 on Build GRCh38
rs373324806 Chr.1: 155287263-155287263 on Build GRCh38
rs370942048 Chr.1: 155287350-155287350 on Build GRCh38
rs751035994 Chr.1: 155287295-155287295 on Build GRCh38
rs1012383857 Chr.1: 155287292-155287292 on Build GRCh38
rs139040609 Chr.1: 155287140-155287140 on Build GRCh38
rs757738076 Chr.1: 155287172-155287172 on Build GRCh38
rs922447856 Chr.1: 155287188-155287188 on Build GRCh38
rs149894199 Chr.1: 155287210-155287210 on Build GRCh38
rs764468982 Chr.1: 155287157-155287157 on Build GRCh38
rs369007284 Chr.1: 155287196-155287196 on Build GRCh38
rs376249401 Chr.1: 155287337-155287337 on Build GRCh38
rs775699845 Chr.1: 155287257-155287257 on Build GRCh38
rs201015446 Chr.1: 155287315-155287315 on Build GRCh38
rs770752686 Chr.1: 155287323-155287323 on Build GRCh38
rs769425868 Chr.1: 155287242-155287242 on Build GRCh38
rs911101122 Chr.1: 155287228-155287228 on Build GRCh38
rs763412553 Chr.1: 155287347-155287347 on Build GRCh38
rs758765081 Chr.1: 155287317-155287317 on Build GRCh38
rs746576195 Chr.1: 155287214-155287214 on Build GRCh38
rs369702343 Chr.1: 155287307-155287307 on Build GRCh38
rs777426562 Chr.1: 155287321-155287321 on Build GRCh38
rs762121064 Chr.1: 155287279-155287279 on Build GRCh38
rs41264937 Chr.1: 155287296-155287296 on Build GRCh38
rs764382725 Chr.1: 155287258-155287258 on Build GRCh38
rs774471370 Chr.1: 155287331-155287331 on Build GRCh38
rs146467190 Chr.1: 155287227-155287227 on Build GRCh38
rs376457661 Chr.1: 155287159-155287159 on Build GRCh38
rs760975472 Chr.1: 155287148-155287148 on Build GRCh38
rs748372125 Chr.1: 155287128-155287129 on Build GRCh38
rs1056120232 Chr.1: 155287287-155287287 on Build GRCh38
rs200927035 Chr.1: 155287286-155287286 on Build GRCh38
rs745834184 Chr.1: 155287332-155287332 on Build GRCh38

View Full Product Details