Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89578776-89579015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
-
UniGene
Hs.461775
Gene Chromosome Location
Chr.16: 89575768-89597246 on Build GRCh38
Transcripts
NM_014427.4 NM_153636.2 XM_011523000.1 XM_011523001.2 XM_017023138.1 XM_017023139.1 XM_017023140.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063621 c.385G>A p.V129M Chr.16: 89578884-89578884 on Build GRCh38
4841032 c.384C>T p.V128V Chr.16: 89578883-89578883 on Build GRCh38
5626698 c.398C>G p.S133C Chr.16: 89578897-89578897 on Build GRCh38
7211673 c.421G>A p.G141R Chr.16: 89578920-89578920 on Build GRCh38
4674397 c.445T>C p.C149R Chr.16: 89578944-89578944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369250260 Chr.16: 89578807-89578807 on Build GRCh38
rs373648106 Chr.16: 89578951-89578951 on Build GRCh38
rs763008448 Chr.16: 89578967-89578967 on Build GRCh38
rs374128987 Chr.16: 89578906-89578908 on Build GRCh38
rs368043912 Chr.16: 89578977-89578977 on Build GRCh38
rs146051306 Chr.16: 89578978-89578978 on Build GRCh38
rs778904372 Chr.16: 89578818-89578818 on Build GRCh38
rs78176582 Chr.16: 89578913-89578913 on Build GRCh38
rs987045624 Chr.16: 89578846-89578846 on Build GRCh38
rs534210956 Chr.16: 89578975-89578975 on Build GRCh38
rs369025222 Chr.16: 89578852-89578852 on Build GRCh38
rs765246853 Chr.16: 89578914-89578914 on Build GRCh38
rs78990556 Chr.16: 89578960-89578960 on Build GRCh38
rs142856114 Chr.16: 89578976-89578976 on Build GRCh38
rs766533546 Chr.16: 89578918-89578918 on Build GRCh38
rs755329950 Chr.16: 89578921-89578921 on Build GRCh38
rs761700586 Chr.16: 89578957-89578957 on Build GRCh38
rs773782633 Chr.16: 89578877-89578877 on Build GRCh38
rs77799181 Chr.16: 89578898-89578898 on Build GRCh38
rs374006079 Chr.16: 89578917-89578917 on Build GRCh38
rs750162314 Chr.16: 89578981-89578983 on Build GRCh38
rs945303270 Chr.16: 89578853-89578853 on Build GRCh38
rs1037651104 Chr.16: 89578925-89578925 on Build GRCh38
rs781714858 Chr.16: 89578860-89578860 on Build GRCh38
rs377581262 Chr.16: 89578845-89578845 on Build GRCh38
rs377398671 Chr.16: 89578902-89578902 on Build GRCh38
rs146919277 Chr.16: 89578859-89578859 on Build GRCh38
rs746701832 Chr.16: 89578950-89578950 on Build GRCh38
rs780286654 Chr.16: 89578835-89578835 on Build GRCh38
rs149812493 Chr.16: 89578952-89578952 on Build GRCh38
rs751839737 Chr.16: 89578920-89578920 on Build GRCh38
rs760485243 Chr.16: 89578899-89578899 on Build GRCh38
rs745389352 Chr.16: 89578884-89578884 on Build GRCh38
rs373180490 Chr.16: 89578827-89578827 on Build GRCh38
rs747309371 Chr.16: 89578837-89578837 on Build GRCh38
rs757356211 Chr.16: 89578814-89578814 on Build GRCh38
rs974708225 Chr.16: 89578801-89578801 on Build GRCh38
rs561379517 Chr.16: 89578807-89578808 on Build GRCh38
rs750640063 Chr.16: 89578823-89578823 on Build GRCh38
rs753308974 Chr.16: 89578831-89578832 on Build GRCh38
rs1043686901 Chr.16: 89578855-89578855 on Build GRCh38
rs933380879 Chr.16: 89578839-89578839 on Build GRCh38
rs753780727 Chr.16: 89578808-89578808 on Build GRCh38
rs748667922 Chr.16: 89578862-89578862 on Build GRCh38
rs778371454 Chr.16: 89578935-89578935 on Build GRCh38
rs141048497 Chr.16: 89578929-89578929 on Build GRCh38
rs527294284 Chr.16: 89578883-89578883 on Build GRCh38
rs775280836 Chr.16: 89578949-89578949 on Build GRCh38
rs903757169 Chr.16: 89578870-89578870 on Build GRCh38
rs9944362 Chr.16: 89578937-89578937 on Build GRCh38
rs776602589 Chr.16: 89578907-89578907 on Build GRCh38
rs771806328 Chr.16: 89578889-89578889 on Build GRCh38
rs374165970 Chr.16: 89578868-89578868 on Build GRCh38
rs148102686 Chr.16: 89578966-89578966 on Build GRCh38
rs146913973 Chr.16: 89578928-89578928 on Build GRCh38
rs761465540 Chr.16: 89578873-89578873 on Build GRCh38
rs370192350 Chr.16: 89578940-89578940 on Build GRCh38

View Full Product Details