Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1043395-1043628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA7
Entrez Gene ID
10347
Gene Name
ATP binding cassette subfamily A member 7
Gene Aliases
ABCA-SSN, ABCX, AD9
UniGene
Hs.134514
Gene Chromosome Location
Chr.19: 1040103-1065572 on Build GRCh38
Transcripts
NM_019112.3 XM_006722616.1 XM_006722617.2 XM_011527628.2 XM_011527629.1 XM_011527630.1 XM_011527631.1 XM_011527632.1 XM_011527633.2 XM_011527634.1 XM_011527635.1 XM_017026142.1 XM_017026143.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5790318 c.930G>C p.Q310H Chr.19: 1043473-1043473 on Build GRCh38
6149778 c.924G>C p.M308I Chr.19: 1043467-1043467 on Build GRCh38
41089 c.856C>T p.R286C Chr.19: 1043399-1043399 on Build GRCh38
84457 c.861G>A p.L287L Chr.19: 1043404-1043404 on Build GRCh38
3149150 c.930+2T>A p.? Chr.19: 1043475-1043475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557177777 Chr.19: 1043569-1043569 on Build GRCh38
rs758750382 Chr.19: 1043495-1043495 on Build GRCh38
rs919012985 Chr.19: 1043554-1043554 on Build GRCh38
rs751875563 Chr.19: 1043497-1043497 on Build GRCh38
rs967495190 Chr.19: 1043476-1043476 on Build GRCh38
rs368234738 Chr.19: 1043515-1043515 on Build GRCh38
rs773931881 Chr.19: 1043422-1043422 on Build GRCh38
rs760140441 Chr.19: 1043465-1043465 on Build GRCh38
rs545443027 Chr.19: 1043559-1043559 on Build GRCh38
rs1028956739 Chr.19: 1043448-1043448 on Build GRCh38
rs749779324 Chr.19: 1043504-1043505 on Build GRCh38
rs781511863 Chr.19: 1043507-1043507 on Build GRCh38
rs749959049 Chr.19: 1043456-1043456 on Build GRCh38
rs3752230 Chr.19: 1043576-1043576 on Build GRCh38
rs561550240 Chr.19: 1043597-1043597 on Build GRCh38
rs913301046 Chr.19: 1043473-1043473 on Build GRCh38
rs377652801 Chr.19: 1043590-1043590 on Build GRCh38
rs1042997911 Chr.19: 1043596-1043596 on Build GRCh38
rs767063313 Chr.19: 1043450-1043450 on Build GRCh38
rs867983999 Chr.19: 1043467-1043467 on Build GRCh38
rs765669177 Chr.19: 1043471-1043471 on Build GRCh38
rs757527196 Chr.19: 1043505-1043505 on Build GRCh38
rs778216019 Chr.19: 1043552-1043552 on Build GRCh38
rs145839896 Chr.19: 1043419-1043419 on Build GRCh38
rs977909247 Chr.19: 1043494-1043494 on Build GRCh38
rs753224215 Chr.19: 1043551-1043551 on Build GRCh38
rs916221115 Chr.19: 1043544-1043544 on Build GRCh38
rs578025097 Chr.19: 1043529-1043529 on Build GRCh38
rs111762673 Chr.19: 1043598-1043598 on Build GRCh38
rs761450156 Chr.19: 1043437-1043437 on Build GRCh38
rs987497758 Chr.19: 1043459-1043459 on Build GRCh38
rs369264049 Chr.19: 1043447-1043447 on Build GRCh38
rs753193022 Chr.19: 1043488-1043488 on Build GRCh38
rs576688236 Chr.19: 1043542-1043543 on Build GRCh38
rs1037840176 Chr.19: 1043573-1043573 on Build GRCh38
rs936652217 Chr.19: 1043599-1043599 on Build GRCh38
rs908841827 Chr.19: 1043478-1043478 on Build GRCh38
rs756148927 Chr.19: 1043525-1043525 on Build GRCh38
rs768438223 Chr.19: 1043420-1043420 on Build GRCh38
rs78017624 Chr.19: 1043416-1043416 on Build GRCh38
rs933091199 Chr.19: 1043558-1043558 on Build GRCh38

View Full Product Details