Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 49280790-49281063 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLN1
Entrez Gene ID
869
Gene Name
cerebellin 1 precursor
Gene Aliases
-
UniGene
Hs.458423
Gene Chromosome Location
Chr.16: 49277917-49281831 on Build GRCh38
Transcripts
NM_004352.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1740234 c.373C>A p.Q125K Chr.16: 49280934-49280934 on Build GRCh38
5984767 c.311T>A p.F104Y Chr.16: 49280996-49280996 on Build GRCh38
471771 c.267A>T p.V89V Chr.16: 49281040-49281040 on Build GRCh38
471770 c.368A>G p.N123S Chr.16: 49280939-49280939 on Build GRCh38
5984766 c.320C>A p.P107Q Chr.16: 49280987-49280987 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746765361 Chr.16: 49280910-49280910 on Build GRCh38
rs759636587 Chr.16: 49280882-49280882 on Build GRCh38
rs199831998 Chr.16: 49280977-49280977 on Build GRCh38
rs753634748 Chr.16: 49280880-49280881 on Build GRCh38
rs34259117 Chr.16: 49280881-49280881 on Build GRCh38
rs145539759 Chr.16: 49281005-49281005 on Build GRCh38
rs763088041 Chr.16: 49280949-49280949 on Build GRCh38
rs775490210 Chr.16: 49280941-49280941 on Build GRCh38
rs374239115 Chr.16: 49280901-49280901 on Build GRCh38
rs1026635956 Chr.16: 49280850-49280850 on Build GRCh38
rs750848489 Chr.16: 49280894-49280894 on Build GRCh38
rs777543103 Chr.16: 49281029-49281029 on Build GRCh38
rs757040561 Chr.16: 49280913-49280913 on Build GRCh38
rs758549268 Chr.16: 49280886-49280886 on Build GRCh38
rs776505732 Chr.16: 49280916-49280916 on Build GRCh38
rs1032237610 Chr.16: 49281030-49281030 on Build GRCh38
rs371075663 Chr.16: 49280885-49280885 on Build GRCh38
rs999285924 Chr.16: 49281028-49281028 on Build GRCh38
rs751829858 Chr.16: 49280978-49280978 on Build GRCh38
rs61735589 Chr.16: 49280926-49280926 on Build GRCh38
rs539174314 Chr.16: 49280860-49280860 on Build GRCh38
rs762047557 Chr.16: 49280983-49280983 on Build GRCh38
rs1009430172 Chr.16: 49280988-49280988 on Build GRCh38
rs771841084 Chr.16: 49280908-49280908 on Build GRCh38
rs780294951 Chr.16: 49280969-49280969 on Build GRCh38
rs763033206 Chr.16: 49280952-49280952 on Build GRCh38
rs776471454 Chr.16: 49280881-49280881 on Build GRCh38
rs553230545 Chr.16: 49280817-49280817 on Build GRCh38
rs778221974 Chr.16: 49280900-49280900 on Build GRCh38
rs973830898 Chr.16: 49280821-49280821 on Build GRCh38
rs772201897 Chr.16: 49280925-49280925 on Build GRCh38
rs373126504 Chr.16: 49280827-49280827 on Build GRCh38
rs369621741 Chr.16: 49280867-49280867 on Build GRCh38
rs757659587 Chr.16: 49280890-49280890 on Build GRCh38
rs199747906 Chr.16: 49281012-49281012 on Build GRCh38
rs916229042 Chr.16: 49280846-49280846 on Build GRCh38
rs769773911 Chr.16: 49280929-49280929 on Build GRCh38
rs200242292 Chr.16: 49280879-49280879 on Build GRCh38
rs756678714 Chr.16: 49280896-49280896 on Build GRCh38
rs542340155 Chr.16: 49280858-49280858 on Build GRCh38
rs770677107 Chr.16: 49280915-49280915 on Build GRCh38
rs375377776 Chr.16: 49280889-49280889 on Build GRCh38
rs767736995 Chr.16: 49280994-49280994 on Build GRCh38
rs536521268 Chr.16: 49280811-49280811 on Build GRCh38
rs765737956 Chr.16: 49280852-49280852 on Build GRCh38
rs750807402 Chr.16: 49280995-49280995 on Build GRCh38

View Full Product Details