Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28758384-28758629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222B
Entrez Gene ID
55731
Gene Name
family with sequence similarity 222 member B
Gene Aliases
C17orf63
UniGene
Hs.564533
Gene Chromosome Location
Chr.17: 28755978-28855036 on Build GRCh38
Transcripts
NM_001077498.2 NM_001288631.1 NM_001288632.1 NM_001288633.1 NM_001288634.1 NM_001288635.1 NM_001288636.1 NM_001288637.1 NM_001288638.1 NM_001288639.1 NM_001288640.1 NM_018182.3 XM_011524986.2 XM_011524987.1 XM_011524988.2 XM_017024831.1 XM_017024832.1 XM_017024833.1 XM_017024834.1 XM_017024835.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3755360 c.1413T>C p.L471L Chr.17: 28758546-28758546 on Build GRCh38
1198252 c.1463C>T p.A488V Chr.17: 28758496-28758496 on Build GRCh38
1381756 c.1341T>C p.G447G Chr.17: 28758618-28758618 on Build GRCh38
7252515 c.1540G>T p.V514L Chr.17: 28758419-28758419 on Build GRCh38
4065069 c.1499G>T p.G500V Chr.17: 28758460-28758460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780716742 Chr.17: 28758453-28758453 on Build GRCh38
rs781380484 Chr.17: 28758539-28758539 on Build GRCh38
rs747967103 Chr.17: 28758567-28758567 on Build GRCh38
rs183643656 Chr.17: 28758561-28758561 on Build GRCh38
rs780828524 Chr.17: 28758530-28758530 on Build GRCh38
rs200850557 Chr.17: 28758549-28758549 on Build GRCh38
rs376696824 Chr.17: 28758537-28758537 on Build GRCh38
rs966908668 Chr.17: 28758428-28758428 on Build GRCh38
rs376632053 Chr.17: 28758472-28758472 on Build GRCh38
rs769746665 Chr.17: 28758406-28758406 on Build GRCh38
rs747492416 Chr.17: 28758421-28758421 on Build GRCh38
rs2043031 Chr.17: 28758546-28758546 on Build GRCh38
rs767143093 Chr.17: 28758605-28758605 on Build GRCh38
rs559809309 Chr.17: 28758603-28758603 on Build GRCh38
rs762089934 Chr.17: 28758485-28758485 on Build GRCh38
rs368737498 Chr.17: 28758522-28758522 on Build GRCh38
rs777468977 Chr.17: 28758457-28758457 on Build GRCh38
rs191008863 Chr.17: 28758525-28758525 on Build GRCh38
rs770559786 Chr.17: 28758574-28758574 on Build GRCh38
rs760820416 Chr.17: 28758575-28758575 on Build GRCh38
rs1022464147 Chr.17: 28758425-28758425 on Build GRCh38
rs578011567 Chr.17: 28758606-28758606 on Build GRCh38
rs759919180 Chr.17: 28758591-28758591 on Build GRCh38
rs200556838 Chr.17: 28758493-28758493 on Build GRCh38
rs973825858 Chr.17: 28758512-28758512 on Build GRCh38
rs201468014 Chr.17: 28758456-28758456 on Build GRCh38
rs762799818 Chr.17: 28758474-28758474 on Build GRCh38
rs774604833 Chr.17: 28758483-28758483 on Build GRCh38
rs918415763 Chr.17: 28758517-28758517 on Build GRCh38
rs201079160 Chr.17: 28758473-28758473 on Build GRCh38
rs760838516 Chr.17: 28758504-28758504 on Build GRCh38
rs371941436 Chr.17: 28758462-28758462 on Build GRCh38
rs369233652 Chr.17: 28758449-28758449 on Build GRCh38
rs760200003 Chr.17: 28758497-28758498 on Build GRCh38
rs746688996 Chr.17: 28758461-28758461 on Build GRCh38
rs536177109 Chr.17: 28758486-28758486 on Build GRCh38
rs912500113 Chr.17: 28758557-28758557 on Build GRCh38
rs973211339 Chr.17: 28758550-28758550 on Build GRCh38
rs750565072 Chr.17: 28758446-28758446 on Build GRCh38
rs879185863 Chr.17: 28758509-28758509 on Build GRCh38
rs754360655 Chr.17: 28758519-28758519 on Build GRCh38
rs373643100 Chr.17: 28758538-28758538 on Build GRCh38
rs751791753 Chr.17: 28758433-28758433 on Build GRCh38
rs756846462 Chr.17: 28758527-28758527 on Build GRCh38
rs775298480 Chr.17: 28758418-28758418 on Build GRCh38
rs755347743 Chr.17: 28758520-28758520 on Build GRCh38
rs762661365 Chr.17: 28758419-28758419 on Build GRCh38
rs749227698 Chr.17: 28758545-28758545 on Build GRCh38
rs758829587 Chr.17: 28758604-28758604 on Build GRCh38
rs766984073 Chr.17: 28758511-28758511 on Build GRCh38
rs767748553 Chr.17: 28758439-28758439 on Build GRCh38
rs772351801 Chr.17: 28758570-28758570 on Build GRCh38
rs768387901 Chr.17: 28758476-28758476 on Build GRCh38
rs773970940 Chr.17: 28758556-28758556 on Build GRCh38
rs777163802 Chr.17: 28758587-28758587 on Build GRCh38
rs754161787 Chr.17: 28758455-28758455 on Build GRCh38
rs775439503 Chr.17: 28758422-28758423 on Build GRCh38
rs766417370 Chr.17: 28758579-28758579 on Build GRCh38

View Full Product Details