Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42263244-42263487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCK
Entrez Gene ID
885
Gene Name
cholecystokinin
Gene Aliases
-
UniGene
Hs.458426
Gene Chromosome Location
Chr.3: 42257824-42266195 on Build GRCh38
Transcripts
NM_000729.5 NM_001174138.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6544381 c.182T>A p.L61Q Chr.3: 42263449-42263449 on Build GRCh38
6622787 c.209G>A p.R70Q Chr.3: 42263422-42263422 on Build GRCh38
3594039 c.146G>A p.R49K Chr.3: 42263485-42263485 on Build GRCh38
371138 c.157G>T p.E53* Chr.3: 42263474-42263474 on Build GRCh38
5864527 c.151G>A p.D51N Chr.3: 42263480-42263480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772953274 Chr.3: 42263465-42263465 on Build GRCh38
rs761668085 Chr.3: 42263338-42263338 on Build GRCh38
rs760723548 Chr.3: 42263374-42263374 on Build GRCh38
rs1002619912 Chr.3: 42263408-42263408 on Build GRCh38
rs779667586 Chr.3: 42263298-42263298 on Build GRCh38
rs769114768 Chr.3: 42263404-42263406 on Build GRCh38
rs757628160 Chr.3: 42263279-42263279 on Build GRCh38
rs779421458 Chr.3: 42263283-42263283 on Build GRCh38
rs754217037 Chr.3: 42263274-42263274 on Build GRCh38
rs568959598 Chr.3: 42263299-42263299 on Build GRCh38
rs771463876 Chr.3: 42263269-42263269 on Build GRCh38
rs200700917 Chr.3: 42263436-42263436 on Build GRCh38
rs747698254 Chr.3: 42263456-42263456 on Build GRCh38
rs773004767 Chr.3: 42263341-42263341 on Build GRCh38
rs765774285 Chr.3: 42263416-42263416 on Build GRCh38
rs754811527 Chr.3: 42263452-42263452 on Build GRCh38
rs376530573 Chr.3: 42263369-42263369 on Build GRCh38
rs199508364 Chr.3: 42263407-42263407 on Build GRCh38
rs1056444222 Chr.3: 42263420-42263420 on Build GRCh38
rs780757604 Chr.3: 42263415-42263415 on Build GRCh38
rs746315394 Chr.3: 42263291-42263291 on Build GRCh38
rs1868522 Chr.3: 42263292-42263292 on Build GRCh38
rs758940296 Chr.3: 42263412-42263412 on Build GRCh38
rs753940802 Chr.3: 42263381-42263381 on Build GRCh38
rs538129810 Chr.3: 42263323-42263323 on Build GRCh38
rs941522669 Chr.3: 42263324-42263324 on Build GRCh38
rs913759095 Chr.3: 42263290-42263290 on Build GRCh38
rs370394067 Chr.3: 42263375-42263375 on Build GRCh38
rs764371425 Chr.3: 42263379-42263379 on Build GRCh38
rs554073050 Chr.3: 42263460-42263460 on Build GRCh38
rs776208885 Chr.3: 42263329-42263329 on Build GRCh38
rs1038570113 Chr.3: 42263339-42263339 on Build GRCh38
rs1052120845 Chr.3: 42263282-42263282 on Build GRCh38
rs757509761 Chr.3: 42263398-42263398 on Build GRCh38
rs199531570 Chr.3: 42263399-42263399 on Build GRCh38
rs760294937 Chr.3: 42263349-42263349 on Build GRCh38
rs766337026 Chr.3: 42263353-42263353 on Build GRCh38
rs768121796 Chr.3: 42263454-42263454 on Build GRCh38

View Full Product Details