Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33278360-33278595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT4
Entrez Gene ID
8705
Gene Name
beta-1,3-galactosyltransferase 4
Gene Aliases
BETA3GALT4, GALT2, GALT4
UniGene
Hs.534375
Gene Chromosome Location
Chr.6: 33277140-33278825 on Build GRCh38
Transcripts
NM_003782.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
159307 c.1124G>T p.W375L Chr.6: 33278543-33278543 on Build GRCh38
6609231 c.1051delG p.E352fs*22 Chr.6: 33278470-33278470 on Build GRCh38
6440516 c.953C>T p.P318L Chr.6: 33278372-33278372 on Build GRCh38
3873133 c.1064C>T p.A355V Chr.6: 33278483-33278483 on Build GRCh38
4654094 c.999G>A p.R333R Chr.6: 33278418-33278418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767249550 Chr.6: 33278531-33278531 on Build GRCh38
rs777889789 Chr.6: 33278575-33278575 on Build GRCh38
rs559571289 Chr.6: 33278520-33278520 on Build GRCh38
rs763301300 Chr.6: 33278502-33278502 on Build GRCh38
rs2231261 Chr.6: 33278415-33278415 on Build GRCh38
rs770000033 Chr.6: 33278487-33278487 on Build GRCh38
rs753821848 Chr.6: 33278553-33278553 on Build GRCh38
rs199514813 Chr.6: 33278568-33278568 on Build GRCh38
rs1018901534 Chr.6: 33278413-33278413 on Build GRCh38
rs369629453 Chr.6: 33278470-33278470 on Build GRCh38
rs771117789 Chr.6: 33278473-33278473 on Build GRCh38
rs756963911 Chr.6: 33278405-33278405 on Build GRCh38
rs377704128 Chr.6: 33278380-33278380 on Build GRCh38
rs772101657 Chr.6: 33278461-33278471 on Build GRCh38
rs374919836 Chr.6: 33278548-33278548 on Build GRCh38
rs146942820 Chr.6: 33278417-33278417 on Build GRCh38
rs763725350 Chr.6: 33278506-33278506 on Build GRCh38
rs906298019 Chr.6: 33278438-33278438 on Build GRCh38
rs143166422 Chr.6: 33278484-33278484 on Build GRCh38
rs149120716 Chr.6: 33278483-33278483 on Build GRCh38
rs780072568 Chr.6: 33278458-33278458 on Build GRCh38
rs763454153 Chr.6: 33278381-33278381 on Build GRCh38
rs766034930 Chr.6: 33278549-33278549 on Build GRCh38
rs72654493 Chr.6: 33278495-33278495 on Build GRCh38
rs4713616 Chr.6: 33278469-33278469 on Build GRCh38
rs1008790390 Chr.6: 33278396-33278396 on Build GRCh38
rs748237478 Chr.6: 33278411-33278411 on Build GRCh38
rs749134945 Chr.6: 33278463-33278463 on Build GRCh38
rs761609610 Chr.6: 33278524-33278524 on Build GRCh38
rs756046614 Chr.6: 33278453-33278453 on Build GRCh38
rs776077315 Chr.6: 33278490-33278490 on Build GRCh38
rs1011232859 Chr.6: 33278558-33278558 on Build GRCh38
rs754869913 Chr.6: 33278562-33278562 on Build GRCh38
rs755960143 Chr.6: 33278537-33278537 on Build GRCh38
rs993522064 Chr.6: 33278465-33278465 on Build GRCh38
rs751260597 Chr.6: 33278390-33278390 on Build GRCh38
rs750057937 Chr.6: 33278533-33278533 on Build GRCh38

View Full Product Details