Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 74522693-74522948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DQX1
Entrez Gene ID
165545
Gene Name
DEAQ-box RNA dependent ATPase 1
Gene Aliases
-
UniGene
Hs.191705
Gene Chromosome Location
Chr.2: 74518131-74526281 on Build GRCh38
Transcripts
NM_133637.2 XM_011532644.1 XM_011532645.1 XM_011532646.2 XM_017003520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4583649 c.915A>G p.A305A Chr.2: 74522890-74522890 on Build GRCh38
4679344 c.1376C>G p.S459* Chr.2: 74522699-74522699 on Build GRCh38
6528363 c.998C>G p.A333G Chr.2: 74522723-74522723 on Build GRCh38
4679346 c.1255_1256insA p.R419fs*36 Chr.2: 74522903-74522904 on Build GRCh38
6564173 c.865G>A p.E289K Chr.2: 74522940-74522940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757402658 Chr.2: 74522859-74522859 on Build GRCh38
rs750713785 Chr.2: 74522878-74522878 on Build GRCh38
rs1024921717 Chr.2: 74522740-74522740 on Build GRCh38
rs760402229 Chr.2: 74522924-74522924 on Build GRCh38
rs974613002 Chr.2: 74522908-74522908 on Build GRCh38
rs771580472 Chr.2: 74522910-74522910 on Build GRCh38
rs757569116 Chr.2: 74522715-74522715 on Build GRCh38
rs774069999 Chr.2: 74522789-74522789 on Build GRCh38
rs781518203 Chr.2: 74522866-74522866 on Build GRCh38
rs781400929 Chr.2: 74522731-74522731 on Build GRCh38
rs780651175 Chr.2: 74522887-74522887 on Build GRCh38
rs751550844 Chr.2: 74522804-74522805 on Build GRCh38
rs770393937 Chr.2: 74522775-74522775 on Build GRCh38
rs747733841 Chr.2: 74522904-74522904 on Build GRCh38
rs772848624 Chr.2: 74522802-74522802 on Build GRCh38
rs141416881 Chr.2: 74522882-74522882 on Build GRCh38
rs758410172 Chr.2: 74522824-74522824 on Build GRCh38
rs760545850 Chr.2: 74522804-74522804 on Build GRCh38
rs780082894 Chr.2: 74522797-74522803 on Build GRCh38
rs761737636 Chr.2: 74522897-74522898 on Build GRCh38
rs190151140 Chr.2: 74522912-74522912 on Build GRCh38
rs753784679 Chr.2: 74522822-74522822 on Build GRCh38
rs200086801 Chr.2: 74522911-74522911 on Build GRCh38
rs748760181 Chr.2: 74522787-74522787 on Build GRCh38
rs749806738 Chr.2: 74522890-74522890 on Build GRCh38
rs768284559 Chr.2: 74522788-74522788 on Build GRCh38
rs766253248 Chr.2: 74522814-74522814 on Build GRCh38
rs760159963 Chr.2: 74522785-74522786 on Build GRCh38
rs370443282 Chr.2: 74522827-74522827 on Build GRCh38
rs916778512 Chr.2: 74522771-74522771 on Build GRCh38
rs1052830647 Chr.2: 74522841-74522841 on Build GRCh38
rs114872355 Chr.2: 74522793-74522793 on Build GRCh38
rs550290623 Chr.2: 74522794-74522794 on Build GRCh38
rs776174287 Chr.2: 74522780-74522780 on Build GRCh38
rs201673170 Chr.2: 74522723-74522723 on Build GRCh38
rs767765043 Chr.2: 74522839-74522839 on Build GRCh38
rs778264602 Chr.2: 74522897-74522897 on Build GRCh38
rs561388327 Chr.2: 74522738-74522738 on Build GRCh38
rs769342695 Chr.2: 74522895-74522895 on Build GRCh38

View Full Product Details