Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18177860-18178085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI30
Entrez Gene ID
10437
Gene Name
IFI30, lysosomal thiol reductase
Gene Aliases
GILT, IFI-30, IP-30, IP30
UniGene
Hs.14623
Gene Chromosome Location
Chr.19: 18173780-18178124 on Build GRCh38
Transcripts
NM_006332.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4830472 c.721A>G p.T241A Chr.19: 18177879-18177879 on Build GRCh38
4750644 c.750G>C p.K250N Chr.19: 18177908-18177908 on Build GRCh38
4506174 c.712C>T p.P238S Chr.19: 18177870-18177870 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1027805866 Chr.19: 18177943-18177943 on Build GRCh38
rs751790931 Chr.19: 18177944-18177944 on Build GRCh38
rs191513645 Chr.19: 18177911-18177911 on Build GRCh38
rs575384663 Chr.19: 18178030-18178030 on Build GRCh38
rs988546380 Chr.19: 18178002-18178002 on Build GRCh38
rs781380689 Chr.19: 18177883-18177883 on Build GRCh38
rs551748369 Chr.19: 18177917-18177917 on Build GRCh38
rs746091568 Chr.19: 18177891-18177891 on Build GRCh38
rs945955468 Chr.19: 18178016-18178016 on Build GRCh38
rs1042896295 Chr.19: 18178040-18178040 on Build GRCh38
rs566703314 Chr.19: 18177989-18177989 on Build GRCh38
rs755323609 Chr.19: 18177978-18177978 on Build GRCh38
rs200628484 Chr.19: 18177880-18177880 on Build GRCh38
rs2921 Chr.19: 18178000-18178000 on Build GRCh38
rs774342582 Chr.19: 18177926-18177926 on Build GRCh38
rs15953 Chr.19: 18177945-18177945 on Build GRCh38
rs77198486 Chr.19: 18177986-18177986 on Build GRCh38
rs555213418 Chr.19: 18178010-18178010 on Build GRCh38
rs184164860 Chr.19: 18177957-18177957 on Build GRCh38
rs539760169 Chr.19: 18177933-18177933 on Build GRCh38
rs571503439 Chr.19: 18177927-18177927 on Build GRCh38
rs15057 Chr.19: 18178012-18178012 on Build GRCh38
rs773175557 Chr.19: 18177936-18177936 on Build GRCh38
rs3185630 Chr.19: 18178055-18178055 on Build GRCh38
rs868346112 Chr.19: 18177916-18177916 on Build GRCh38

View Full Product Details