Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 37011811-37012036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD2
Entrez Gene ID
221472
Gene Name
FYVE, RhoGEF and PH domain containing 2
Gene Aliases
ZFYVE4
UniGene
Hs.509664
Gene Chromosome Location
Chr.6: 37005647-37029072 on Build GRCh38
Transcripts
NM_173558.3 XM_011514372.2 XM_011514373.2 XM_017010429.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6439214 c.523G>A p.D175N Chr.6: 37011850-37011850 on Build GRCh38
186839 c.515G>A p.R172H Chr.6: 37011842-37011842 on Build GRCh38
7209008 c.515G>T p.R172L Chr.6: 37011842-37011842 on Build GRCh38
3430587 c.514C>T p.R172C Chr.6: 37011841-37011841 on Build GRCh38
4841445 c.498C>T p.L166L Chr.6: 37011825-37011825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901127858 Chr.6: 37011922-37011922 on Build GRCh38
rs142895649 Chr.6: 37011839-37011839 on Build GRCh38
rs780269753 Chr.6: 37011850-37011850 on Build GRCh38
rs928215585 Chr.6: 37011986-37011986 on Build GRCh38
rs772545294 Chr.6: 37011895-37011895 on Build GRCh38
rs753995514 Chr.6: 37011871-37011871 on Build GRCh38
rs951478484 Chr.6: 37011868-37011868 on Build GRCh38
rs750711356 Chr.6: 37011842-37011842 on Build GRCh38
rs566830080 Chr.6: 37012014-37012014 on Build GRCh38
rs1020727151 Chr.6: 37011964-37011964 on Build GRCh38
rs961230225 Chr.6: 37011987-37011987 on Build GRCh38
rs1010634636 Chr.6: 37011956-37011956 on Build GRCh38
rs996907775 Chr.6: 37011933-37011933 on Build GRCh38
rs969382422 Chr.6: 37011952-37011952 on Build GRCh38
rs765180579 Chr.6: 37011832-37011832 on Build GRCh38
rs752588458 Chr.6: 37011838-37011838 on Build GRCh38
rs1041321747 Chr.6: 37011917-37011917 on Build GRCh38
rs999033805 Chr.6: 37012009-37012009 on Build GRCh38
rs756298412 Chr.6: 37011849-37011849 on Build GRCh38
rs752321023 Chr.6: 37011927-37011927 on Build GRCh38
rs58465444 Chr.6: 37011893-37011893 on Build GRCh38
rs755071666 Chr.6: 37011872-37011872 on Build GRCh38
rs755658495 Chr.6: 37011954-37011954 on Build GRCh38
rs748645616 Chr.6: 37011894-37011894 on Build GRCh38
rs763769959 Chr.6: 37011841-37011841 on Build GRCh38

View Full Product Details