Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47680953-47681202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLTSCR1
Entrez Gene ID
29998
Gene Name
glioma tumor suppressor candidate region gene 1
Gene Aliases
-
UniGene
Hs.97244
Gene Chromosome Location
Chr.19: 47608196-47703277 on Build GRCh38
Transcripts
NM_015711.3 XM_005258833.4 XM_006723180.3 XM_011526882.2 XM_011526883.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4132201 c.1996C>A p.P666T Chr.19: 47681166-47681166 on Build GRCh38
5033964 c.1819A>C p.T607P Chr.19: 47680989-47680989 on Build GRCh38
7003793 c.1859C>T p.T620M Chr.19: 47681029-47681029 on Build GRCh38
6043291 c.1887C>T p.P629P Chr.19: 47681057-47681057 on Build GRCh38
6558061 c.2023G>T p.E675* Chr.19: 47681193-47681193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957806891 Chr.19: 47680997-47680997 on Build GRCh38
rs576832933 Chr.19: 47681138-47681138 on Build GRCh38
rs938004239 Chr.19: 47681076-47681076 on Build GRCh38
rs1055013721 Chr.19: 47681102-47681102 on Build GRCh38
rs1050506044 Chr.19: 47681136-47681137 on Build GRCh38
rs974857325 Chr.19: 47681015-47681015 on Build GRCh38
rs914028277 Chr.19: 47680976-47680976 on Build GRCh38
rs938571145 Chr.19: 47680989-47680989 on Build GRCh38
rs766990038 Chr.19: 47681122-47681122 on Build GRCh38
rs1013899105 Chr.19: 47681114-47681114 on Build GRCh38
rs1044179899 Chr.19: 47681018-47681018 on Build GRCh38
rs774132000 Chr.19: 47681105-47681113 on Build GRCh38
rs752174033 Chr.19: 47681124-47681124 on Build GRCh38
rs755526837 Chr.19: 47681127-47681127 on Build GRCh38
rs773692212 Chr.19: 47681025-47681025 on Build GRCh38
rs545666349 Chr.19: 47681143-47681143 on Build GRCh38
rs921709788 Chr.19: 47681161-47681161 on Build GRCh38
rs945509256 Chr.19: 47680984-47680984 on Build GRCh38
rs763451029 Chr.19: 47681041-47681041 on Build GRCh38
rs765520797 Chr.19: 47680971-47680971 on Build GRCh38
rs933120401 Chr.19: 47681183-47681183 on Build GRCh38
rs563305326 Chr.19: 47681113-47681113 on Build GRCh38
rs886260303 Chr.19: 47681136-47681136 on Build GRCh38
rs1047572642 Chr.19: 47681135-47681135 on Build GRCh38
rs991453475 Chr.19: 47681069-47681069 on Build GRCh38

View Full Product Details