Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 123382540-123382799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND1A
Entrez Gene ID
57706
Gene Name
DENN domain containing 1A
Gene Aliases
FAM31A, KIAA1608
UniGene
Hs.744995
Gene Chromosome Location
Chr.9: 123379654-123930158 on Build GRCh38
Transcripts
NM_020946.1 XM_005252109.2 XM_005252111.4 XM_006717195.3 XM_011518882.2 XM_011518883.2 XM_011518884.2 XM_011518885.2 XM_011518886.2 XM_017014947.1 XM_017014948.1 XM_017014949.1 XM_017014950.1 XM_017014951.1 XM_017014952.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4386520 c.1885G>A p.G629R Chr.9: 123382610-123382610 on Build GRCh38
1459937 c.1914C>A p.L638L Chr.9: 123382548-123382548 on Build GRCh38
3903957 c.1901G>A p.R634H Chr.9: 123382594-123382594 on Build GRCh38
6642238 c.1935G>A p.P645P Chr.9: 123382560-123382560 on Build GRCh38
3324309 c.1867C>T p.R623C Chr.9: 123382595-123382595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748644744 Chr.9: 123382647-123382647 on Build GRCh38
rs771504403 Chr.9: 123382646-123382646 on Build GRCh38
rs140495915 Chr.9: 123382605-123382605 on Build GRCh38
rs747646467 Chr.9: 123382676-123382677 on Build GRCh38
rs1056888924 Chr.9: 123382636-123382636 on Build GRCh38
rs773887773 Chr.9: 123382564-123382564 on Build GRCh38
rs778444665 Chr.9: 123382674-123382675 on Build GRCh38
rs571261934 Chr.9: 123382725-123382725 on Build GRCh38
rs923373646 Chr.9: 123382773-123382773 on Build GRCh38
rs910615742 Chr.9: 123382751-123382751 on Build GRCh38
rs766643227 Chr.9: 123382659-123382659 on Build GRCh38
rs954847982 Chr.9: 123382734-123382734 on Build GRCh38
rs987625221 Chr.9: 123382750-123382750 on Build GRCh38
rs765585913 Chr.9: 123382579-123382579 on Build GRCh38
rs950938517 Chr.9: 123382746-123382746 on Build GRCh38
rs776559021 Chr.9: 123382758-123382758 on Build GRCh38
rs751689270 Chr.9: 123382594-123382594 on Build GRCh38
rs113183359 Chr.9: 123382724-123382724 on Build GRCh38
rs943357883 Chr.9: 123382591-123382591 on Build GRCh38
rs1037729488 Chr.9: 123382592-123382592 on Build GRCh38
rs763183808 Chr.9: 123382658-123382658 on Build GRCh38
rs202191290 Chr.9: 123382593-123382593 on Build GRCh38
rs527475821 Chr.9: 123382651-123382651 on Build GRCh38
rs1002723210 Chr.9: 123382625-123382625 on Build GRCh38
rs767572605 Chr.9: 123382595-123382595 on Build GRCh38
rs4836919 Chr.9: 123382723-123382723 on Build GRCh38
rs369085843 Chr.9: 123382560-123382560 on Build GRCh38
rs545166170 Chr.9: 123382571-123382571 on Build GRCh38
rs745342804 Chr.9: 123382565-123382565 on Build GRCh38
rs1001921875 Chr.9: 123382691-123382691 on Build GRCh38
rs1033854357 Chr.9: 123382695-123382695 on Build GRCh38
rs372816654 Chr.9: 123382665-123382665 on Build GRCh38
rs531597394 Chr.9: 123382722-123382722 on Build GRCh38
rs756040661 Chr.9: 123382597-123382597 on Build GRCh38
rs770403164 Chr.9: 123382561-123382561 on Build GRCh38
rs763217581 Chr.9: 123382590-123382590 on Build GRCh38
rs371842367 Chr.9: 123382611-123382611 on Build GRCh38
rs770106908 Chr.9: 123382650-123382650 on Build GRCh38
rs774375326 Chr.9: 123382661-123382661 on Build GRCh38
rs910646747 Chr.9: 123382562-123382562 on Build GRCh38
rs985487530 Chr.9: 123382747-123382747 on Build GRCh38
rs745395923 Chr.9: 123382638-123382638 on Build GRCh38
rs974828423 Chr.9: 123382761-123382761 on Build GRCh38
rs768913848 Chr.9: 123382757-123382757 on Build GRCh38
rs549135437 Chr.9: 123382701-123382701 on Build GRCh38
rs561185306 Chr.9: 123382714-123382714 on Build GRCh38
rs1050943508 Chr.9: 123382614-123382614 on Build GRCh38
rs777608404 Chr.9: 123382601-123382601 on Build GRCh38
rs973802438 Chr.9: 123382760-123382760 on Build GRCh38
rs773367767 Chr.9: 123382587-123382587 on Build GRCh38
rs756967284 Chr.9: 123382610-123382610 on Build GRCh38
rs777065780 Chr.9: 123382578-123382578 on Build GRCh38
rs1012586722 Chr.9: 123382676-123382676 on Build GRCh38
rs903723580 Chr.9: 123382671-123382671 on Build GRCh38

View Full Product Details