Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 69699503-69699728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF19
Entrez Gene ID
9965
Gene Name
fibroblast growth factor 19
Gene Aliases
-
UniGene
Hs.249200
Gene Chromosome Location
Chr.11: 69698238-69704338 on Build GRCh38
Transcripts
NM_005117.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6951692 c.379C>T p.R127C Chr.11: 69699534-69699534 on Build GRCh38
6953853 c.410A>G p.E137G Chr.11: 69699503-69699503 on Build GRCh38
6445701 c.402C>T p.Y134Y Chr.11: 69699511-69699511 on Build GRCh38
6928581 c.349G>T p.E117* Chr.11: 69699564-69699564 on Build GRCh38
6663525 c.403C>T p.R135* Chr.11: 69699510-69699510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779547280 Chr.11: 69699585-69699585 on Build GRCh38
rs377419016 Chr.11: 69699546-69699546 on Build GRCh38
rs765440889 Chr.11: 69699566-69699566 on Build GRCh38
rs780574173 Chr.11: 69699602-69699602 on Build GRCh38
rs759680367 Chr.11: 69699547-69699547 on Build GRCh38
rs553838145 Chr.11: 69699626-69699626 on Build GRCh38
rs1022187592 Chr.11: 69699676-69699676 on Build GRCh38
rs1035912586 Chr.11: 69699565-69699565 on Build GRCh38
rs749893226 Chr.11: 69699608-69699608 on Build GRCh38
rs188495602 Chr.11: 69699539-69699539 on Build GRCh38
rs931218641 Chr.11: 69699651-69699651 on Build GRCh38
rs147194982 Chr.11: 69699640-69699640 on Build GRCh38
rs148589027 Chr.11: 69699533-69699533 on Build GRCh38
rs760883667 Chr.11: 69699537-69699537 on Build GRCh38
rs541925556 Chr.11: 69699625-69699625 on Build GRCh38
rs908212906 Chr.11: 69699681-69699681 on Build GRCh38
rs982819739 Chr.11: 69699672-69699672 on Build GRCh38
rs142959398 Chr.11: 69699540-69699540 on Build GRCh38
rs763273506 Chr.11: 69699582-69699582 on Build GRCh38
rs374252033 Chr.11: 69699627-69699627 on Build GRCh38
rs970169758 Chr.11: 69699678-69699678 on Build GRCh38
rs879252494 Chr.11: 69699636-69699636 on Build GRCh38
rs752083948 Chr.11: 69699598-69699598 on Build GRCh38
rs1011780967 Chr.11: 69699661-69699661 on Build GRCh38
rs757744979 Chr.11: 69699600-69699600 on Build GRCh38

View Full Product Details