Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26384972-26385209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPT1
Entrez Gene ID
85465
Gene Name
ethanolaminephosphotransferase 1
Gene Aliases
SELI, SEPI
UniGene
Hs.189073
Gene Chromosome Location
Chr.2: 26346086-26395891 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7247691 c.897T>C p.A299A Chr.2: 26385124-26385124 on Build GRCh38
3580344 c.890G>A p.G297E Chr.2: 26385117-26385117 on Build GRCh38
5923591 c.764C>T p.S255L Chr.2: 26384991-26384991 on Build GRCh38
3910091 c.810C>T p.F270F Chr.2: 26385037-26385037 on Build GRCh38
6502807 c.840A>C p.S280S Chr.2: 26385067-26385067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs186610692 Chr.2: 26385145-26385145 on Build GRCh38
rs541724034 Chr.2: 26385075-26385075 on Build GRCh38
rs376360026 Chr.2: 26385043-26385043 on Build GRCh38
rs763057214 Chr.2: 26385088-26385088 on Build GRCh38
rs775151096 Chr.2: 26385052-26385052 on Build GRCh38
rs952618379 Chr.2: 26385067-26385067 on Build GRCh38
rs543411515 Chr.2: 26385160-26385160 on Build GRCh38
rs752357049 Chr.2: 26385016-26385016 on Build GRCh38
rs767798392 Chr.2: 26385006-26385006 on Build GRCh38
rs767464314 Chr.2: 26385165-26385169 on Build GRCh38
rs748866328 Chr.2: 26385042-26385042 on Build GRCh38
rs748343718 Chr.2: 26385073-26385073 on Build GRCh38
rs752540157 Chr.2: 26385166-26385166 on Build GRCh38
rs372068475 Chr.2: 26385031-26385031 on Build GRCh38
rs763064994 Chr.2: 26385146-26385146 on Build GRCh38
rs767860001 Chr.2: 26385171-26385171 on Build GRCh38
rs934280 Chr.2: 26385124-26385124 on Build GRCh38
rs755731051 Chr.2: 26385025-26385025 on Build GRCh38
rs769967115 Chr.2: 26385074-26385074 on Build GRCh38
rs760755053 Chr.2: 26385174-26385192 on Build GRCh38
rs773435770 Chr.2: 26385145-26385145 on Build GRCh38
rs769942577 Chr.2: 26385139-26385140 on Build GRCh38
rs745637775 Chr.2: 26385050-26385050 on Build GRCh38
rs369548288 Chr.2: 26385051-26385051 on Build GRCh38
rs755417275 Chr.2: 26385004-26385004 on Build GRCh38

View Full Product Details