Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 131275843-131276086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF1
Entrez Gene ID
3547
Gene Name
immunoglobulin superfamily member 1
Gene Aliases
CHTE, IGCD1, IGDC1, INHBP, PGSF2, p120
UniGene
Hs.22111
Gene Chromosome Location
Chr.X: 131273506-131289457 on Build GRCh38
Transcripts
NM_001170961.1 NM_001170962.1 NM_001555.4 XM_011531330.1 XM_011531333.1 XM_011531334.1

SNP Detail

SNP ID Chromosome Location
rs764878217 Chr.X: 131275962-131275962 on Build GRCh38
rs754757866 Chr.X: 131275870-131275870 on Build GRCh38
rs752534920 Chr.X: 131275964-131275964 on Build GRCh38
rs761234734 Chr.X: 131275888-131275888 on Build GRCh38
rs370093373 Chr.X: 131275947-131275947 on Build GRCh38
rs201109191 Chr.X: 131275976-131275976 on Build GRCh38
rs774618524 Chr.X: 131275944-131275944 on Build GRCh38
rs766446436 Chr.X: 131275949-131275949 on Build GRCh38
rs780338952 Chr.X: 131276026-131276026 on Build GRCh38
rs745979788 Chr.X: 131276059-131276059 on Build GRCh38
rs374075974 Chr.X: 131275951-131275951 on Build GRCh38
rs750770924 Chr.X: 131275869-131275869 on Build GRCh38
rs750861873 Chr.X: 131276011-131276011 on Build GRCh38
rs867723820 Chr.X: 131275948-131275948 on Build GRCh38
rs768908371 Chr.X: 131275940-131275940 on Build GRCh38
rs1056672499 Chr.X: 131275877-131275877 on Build GRCh38
rs200115183 Chr.X: 131275952-131275952 on Build GRCh38
rs756639396 Chr.X: 131276013-131276013 on Build GRCh38
rs201754584 Chr.X: 131275991-131275991 on Build GRCh38
rs375506944 Chr.X: 131275922-131275922 on Build GRCh38
rs371951904 Chr.X: 131275992-131275992 on Build GRCh38
rs111999883 Chr.X: 131275881-131275881 on Build GRCh38
rs1053176557 Chr.X: 131276043-131276043 on Build GRCh38
rs192366574 Chr.X: 131276055-131276055 on Build GRCh38
rs1029188801 Chr.X: 131276057-131276057 on Build GRCh38
rs55936630 Chr.X: 131275910-131275910 on Build GRCh38
rs751111728 Chr.X: 131275989-131275989 on Build GRCh38
rs762573543 Chr.X: 131275965-131275965 on Build GRCh38
rs73635985 Chr.X: 131275927-131275927 on Build GRCh38
rs766942342 Chr.X: 131275901-131275901 on Build GRCh38

View Full Product Details