Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32769938-32770167 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1522
Entrez Gene ID
57648
Gene Name
KIAA1522
Gene Aliases
-
UniGene
Hs.591502
Gene Chromosome Location
Chr.1: 32741839-32774970 on Build GRCh38
Transcripts
NM_001198972.1 NM_001198973.1 NM_020888.2 XM_006710791.3 XM_017001916.1 XM_017001917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5846408 c.592C>T p.P198S Chr.1: 32769948-32769948 on Build GRCh38
5952850 c.886A>G p.I296V Chr.1: 32770065-32770065 on Build GRCh38
2078269 c.663G>A p.A221A Chr.1: 32770019-32770019 on Build GRCh38
2078271 c.677G>A p.G226D Chr.1: 32770033-32770033 on Build GRCh38
3943876 c.897C>G p.V299V Chr.1: 32770076-32770076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776206125 Chr.1: 32770050-32770051 on Build GRCh38
rs753297494 Chr.1: 32769981-32769981 on Build GRCh38
rs771185027 Chr.1: 32770089-32770089 on Build GRCh38
rs375691123 Chr.1: 32770080-32770080 on Build GRCh38
rs754751165 Chr.1: 32770135-32770135 on Build GRCh38
rs768088384 Chr.1: 32769977-32769977 on Build GRCh38
rs772554806 Chr.1: 32770101-32770101 on Build GRCh38
rs537613971 Chr.1: 32769982-32769982 on Build GRCh38
rs781167678 Chr.1: 32770142-32770142 on Build GRCh38
rs756778680 Chr.1: 32770047-32770047 on Build GRCh38
rs763291208 Chr.1: 32769963-32769963 on Build GRCh38
rs750060794 Chr.1: 32770121-32770121 on Build GRCh38
rs754022301 Chr.1: 32770018-32770018 on Build GRCh38
rs12730560 Chr.1: 32770050-32770050 on Build GRCh38
rs760954841 Chr.1: 32770107-32770107 on Build GRCh38
rs778546155 Chr.1: 32769994-32769994 on Build GRCh38
rs79767347 Chr.1: 32770040-32770040 on Build GRCh38
rs899372417 Chr.1: 32769970-32769970 on Build GRCh38
rs374708941 Chr.1: 32770034-32770034 on Build GRCh38
rs775914331 Chr.1: 32770102-32770102 on Build GRCh38
rs781023475 Chr.1: 32770072-32770072 on Build GRCh38
rs370646316 Chr.1: 32769960-32769960 on Build GRCh38
rs751904577 Chr.1: 32769967-32769967 on Build GRCh38
rs368418447 Chr.1: 32770091-32770091 on Build GRCh38
rs759654546 Chr.1: 32769959-32769959 on Build GRCh38
rs746399421 Chr.1: 32770012-32770013 on Build GRCh38
rs543606757 Chr.1: 32770116-32770116 on Build GRCh38
rs766721328 Chr.1: 32769964-32769964 on Build GRCh38
rs768222894 Chr.1: 32770013-32770018 on Build GRCh38
rs113474194 Chr.1: 32770059-32770059 on Build GRCh38
rs764526872 Chr.1: 32770043-32770043 on Build GRCh38
rs3737995 Chr.1: 32770025-32770025 on Build GRCh38
rs374723336 Chr.1: 32770000-32770000 on Build GRCh38
rs774736808 Chr.1: 32770023-32770023 on Build GRCh38
rs554356770 Chr.1: 32770049-32770049 on Build GRCh38
rs746791481 Chr.1: 32770065-32770065 on Build GRCh38
rs370384732 Chr.1: 32770009-32770009 on Build GRCh38
rs775858898 Chr.1: 32770037-32770037 on Build GRCh38
rs777049903 Chr.1: 32770111-32770111 on Build GRCh38
rs867084511 Chr.1: 32770138-32770139 on Build GRCh38
rs745609741 Chr.1: 32769996-32769996 on Build GRCh38
rs565146691 Chr.1: 32770117-32770117 on Build GRCh38
rs746161726 Chr.1: 32770094-32770094 on Build GRCh38
rs757986626 Chr.1: 32770127-32770127 on Build GRCh38
rs765950712 Chr.1: 32770128-32770128 on Build GRCh38
rs753326180 Chr.1: 32770046-32770046 on Build GRCh38
rs761774664 Chr.1: 32769955-32769955 on Build GRCh38
rs556325050 Chr.1: 32769997-32769997 on Build GRCh38
rs760010929 Chr.1: 32769968-32769968 on Build GRCh38
rs779955970 Chr.1: 32770060-32770060 on Build GRCh38
rs753423330 Chr.1: 32769991-32769991 on Build GRCh38
rs201266399 Chr.1: 32770071-32770071 on Build GRCh38
rs942343620 Chr.1: 32770081-32770081 on Build GRCh38
rs772246869 Chr.1: 32769993-32769993 on Build GRCh38
rs200423928 Chr.1: 32770108-32770108 on Build GRCh38
rs371267433 Chr.1: 32770010-32770010 on Build GRCh38

View Full Product Details