Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 9025432-9025693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM9B
Entrez Gene ID
171483
Gene Name
family with sequence similarity 9 member B
Gene Aliases
TEX39B
UniGene
Hs.733351
Gene Chromosome Location
Chr.X: 9024232-9197176 on Build GRCh38
Transcripts
NM_205849.3 XM_011545464.2

SNP Detail

SNP ID Chromosome Location
rs745564131 Chr.X: 9025551-9025551 on Build GRCh38
rs192751912 Chr.X: 9025572-9025572 on Build GRCh38
rs148142009 Chr.X: 9025630-9025630 on Build GRCh38
rs984780031 Chr.X: 9025645-9025645 on Build GRCh38
rs774368116 Chr.X: 9025631-9025631 on Build GRCh38
rs371492822 Chr.X: 9025478-9025478 on Build GRCh38
rs910111113 Chr.X: 9025659-9025659 on Build GRCh38
rs761732729 Chr.X: 9025619-9025619 on Build GRCh38
rs776684965 Chr.X: 9025614-9025614 on Build GRCh38
rs767895428 Chr.X: 9025511-9025511 on Build GRCh38
rs201649265 Chr.X: 9025555-9025555 on Build GRCh38
rs765528348 Chr.X: 9025477-9025480 on Build GRCh38
rs200690989 Chr.X: 9025541-9025541 on Build GRCh38
rs746747410 Chr.X: 9025585-9025585 on Build GRCh38
rs374635891 Chr.X: 9025521-9025521 on Build GRCh38
rs143823179 Chr.X: 9025542-9025542 on Build GRCh38
rs759735838 Chr.X: 9025483-9025483 on Build GRCh38
rs368843004 Chr.X: 9025582-9025582 on Build GRCh38
rs751821791 Chr.X: 9025482-9025482 on Build GRCh38
rs753070325 Chr.X: 9025518-9025518 on Build GRCh38
rs748282640 Chr.X: 9025461-9025461 on Build GRCh38
rs763029165 Chr.X: 9025469-9025469 on Build GRCh38
rs959531861 Chr.X: 9025607-9025607 on Build GRCh38
rs779744265 Chr.X: 9025562-9025562 on Build GRCh38
rs372765352 Chr.X: 9025602-9025602 on Build GRCh38
rs778285761 Chr.X: 9025533-9025533 on Build GRCh38
rs756584379 Chr.X: 9025528-9025528 on Build GRCh38
rs758487611 Chr.X: 9025586-9025591 on Build GRCh38
rs752791732 Chr.X: 9025543-9025544 on Build GRCh38
rs778533482 Chr.X: 9025547-9025547 on Build GRCh38

View Full Product Details