Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168379322-168379565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX60L
Entrez Gene ID
91351
Gene Name
DEAD-box helicase 60-like
Gene Aliases
-
UniGene
Hs.535011
Gene Chromosome Location
Chr.4: 168356735-168480514 on Build GRCh38
Transcripts
NM_001012967.2 XM_005263341.4 XM_005263342.2 XM_005263343.2 XM_005263345.4 XM_006714408.2 XM_011532402.2 XM_011532403.2 XM_011532404.1 XM_011532405.1 XM_011532406.1 XM_011532409.2 XM_017008829.1 XM_017008830.1 XM_017008831.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5912060 c.4271C>T p.S1424L Chr.4: 168379455-168379455 on Build GRCh38
5529657 c.4342C>T p.L1448F Chr.4: 168379384-168379384 on Build GRCh38
6099953 c.4268C>T p.A1423V Chr.4: 168379458-168379458 on Build GRCh38
733149 c.4320C>T p.L1440L Chr.4: 168379406-168379406 on Build GRCh38
5018959 c.4244A>T p.N1415I Chr.4: 168379482-168379482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762236190 Chr.4: 168379511-168379511 on Build GRCh38
rs761706772 Chr.4: 168379358-168379358 on Build GRCh38
rs376620813 Chr.4: 168379526-168379526 on Build GRCh38
rs755011338 Chr.4: 168379389-168379389 on Build GRCh38
rs779470398 Chr.4: 168379523-168379525 on Build GRCh38
rs959745035 Chr.4: 168379475-168379475 on Build GRCh38
rs773068004 Chr.4: 168379505-168379505 on Build GRCh38
rs199826168 Chr.4: 168379467-168379467 on Build GRCh38
rs376955320 Chr.4: 168379446-168379446 on Build GRCh38
rs889885382 Chr.4: 168379374-168379374 on Build GRCh38
rs752860577 Chr.4: 168379408-168379408 on Build GRCh38
rs750042433 Chr.4: 168379520-168379520 on Build GRCh38
rs372407110 Chr.4: 168379429-168379429 on Build GRCh38
rs371784309 Chr.4: 168379486-168379486 on Build GRCh38
rs199708273 Chr.4: 168379448-168379448 on Build GRCh38
rs998676338 Chr.4: 168379347-168379347 on Build GRCh38
rs766363640 Chr.4: 168379521-168379521 on Build GRCh38
rs749588282 Chr.4: 168379426-168379426 on Build GRCh38
rs765062773 Chr.4: 168379376-168379376 on Build GRCh38
rs368384460 Chr.4: 168379409-168379409 on Build GRCh38
rs767588674 Chr.4: 168379392-168379392 on Build GRCh38
rs757111143 Chr.4: 168379362-168379362 on Build GRCh38
rs1049866203 Chr.4: 168379373-168379373 on Build GRCh38
rs375382515 Chr.4: 168379346-168379346 on Build GRCh38
rs553888843 Chr.4: 168379523-168379523 on Build GRCh38
rs750395940 Chr.4: 168379377-168379377 on Build GRCh38
rs778407887 Chr.4: 168379507-168379507 on Build GRCh38
rs772433566 Chr.4: 168379519-168379519 on Build GRCh38
rs778126545 Chr.4: 168379417-168379417 on Build GRCh38
rs758076758 Chr.4: 168379521-168379525 on Build GRCh38
rs776284657 Chr.4: 168379354-168379354 on Build GRCh38
rs746378834 Chr.4: 168379434-168379434 on Build GRCh38
rs1022458020 Chr.4: 168379525-168379525 on Build GRCh38
rs200801211 Chr.4: 168379350-168379350 on Build GRCh38
rs539267006 Chr.4: 168379484-168379484 on Build GRCh38
rs760851686 Chr.4: 168379528-168379528 on Build GRCh38
rs771646763 Chr.4: 168379438-168379438 on Build GRCh38

View Full Product Details