Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 127175338-127175599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK1
Entrez Gene ID
1793
Gene Name
dedicator of cytokinesis 1
Gene Aliases
DOCK180, ced5
UniGene
Hs.159195
Gene Chromosome Location
Chr.10: 126905409-127452517 on Build GRCh38
Transcripts
NM_001290223.1 NM_001380.4 XM_011539422.2 XM_011539424.1 XM_017015813.1 XM_017015814.1 XM_017015815.1 XM_017015816.1 NM_001039762.2 XM_005252694.3 XM_017016537.1 XM_017016538.1 XM_017016539.1 XM_017016540.1 XM_017016541.1 XM_017016542.1 XM_017016543.1

Gene Symbol
FAM196A
Entrez Gene ID
642938
Gene Name
family with sequence similarity 196 member A
Gene Aliases
C10orf141
UniGene
Hs.613882
Gene Chromosome Location
Chr.10: 127135426-127196522 on Build GRCh38
Transcripts
NM_001290223.1 NM_001380.4 XM_011539422.2 XM_011539424.1 XM_017015813.1 XM_017015814.1 XM_017015815.1 XM_017015816.1 NM_001039762.2 XM_005252694.3 XM_017016537.1 XM_017016538.1 XM_017016539.1 XM_017016540.1 XM_017016541.1 XM_017016542.1 XM_017016543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6408363 c.895G>A p.A299T Chr.10: 127175501-127175501 on Build GRCh38
684060 c.930G>C p.P310P Chr.10: 127175466-127175466 on Build GRCh38
260790 c.1047C>T p.I349I Chr.10: 127175349-127175349 on Build GRCh38
5469366 c.903G>A p.S301S Chr.10: 127175493-127175493 on Build GRCh38
1346704 c.984G>A p.P328P Chr.10: 127175412-127175412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377729347 Chr.10: 127175440-127175440 on Build GRCh38
rs764156865 Chr.10: 127175538-127175538 on Build GRCh38
rs766379230 Chr.10: 127175534-127175534 on Build GRCh38
rs781714055 Chr.10: 127175438-127175438 on Build GRCh38
rs757623149 Chr.10: 127175505-127175505 on Build GRCh38
rs747882538 Chr.10: 127175529-127175529 on Build GRCh38
rs758739317 Chr.10: 127175428-127175428 on Build GRCh38
rs765252753 Chr.10: 127175489-127175489 on Build GRCh38
rs778371072 Chr.10: 127175577-127175577 on Build GRCh38
rs771469519 Chr.10: 127175379-127175379 on Build GRCh38
rs200373232 Chr.10: 127175511-127175511 on Build GRCh38
rs61752578 Chr.10: 127175382-127175382 on Build GRCh38
rs762755076 Chr.10: 127175384-127175384 on Build GRCh38
rs149479141 Chr.10: 127175537-127175537 on Build GRCh38
rs547642648 Chr.10: 127175554-127175554 on Build GRCh38
rs371917458 Chr.10: 127175548-127175548 on Build GRCh38
rs527749367 Chr.10: 127175409-127175409 on Build GRCh38
rs368886635 Chr.10: 127175547-127175547 on Build GRCh38
rs763819554 Chr.10: 127175400-127175400 on Build GRCh38
rs373397161 Chr.10: 127175466-127175466 on Build GRCh38
rs747451108 Chr.10: 127175430-127175430 on Build GRCh38
rs779209910 Chr.10: 127175422-127175422 on Build GRCh38
rs866018984 Chr.10: 127175492-127175492 on Build GRCh38
rs958344442 Chr.10: 127175501-127175501 on Build GRCh38
rs1027675265 Chr.10: 127175571-127175571 on Build GRCh38
rs773235819 Chr.10: 127175530-127175530 on Build GRCh38
rs767609478 Chr.10: 127175410-127175410 on Build GRCh38
rs767554325 Chr.10: 127175463-127175463 on Build GRCh38
rs143897472 Chr.10: 127175570-127175570 on Build GRCh38
rs748875288 Chr.10: 127175519-127175519 on Build GRCh38
rs144141274 Chr.10: 127175380-127175380 on Build GRCh38
rs150869481 Chr.10: 127175500-127175500 on Build GRCh38
rs779403804 Chr.10: 127175512-127175512 on Build GRCh38
rs753949881 Chr.10: 127175412-127175412 on Build GRCh38
rs982182071 Chr.10: 127175377-127175377 on Build GRCh38
rs137995652 Chr.10: 127175521-127175521 on Build GRCh38
rs768391430 Chr.10: 127175455-127175455 on Build GRCh38
rs751794894 Chr.10: 127175542-127175542 on Build GRCh38
rs754517108 Chr.10: 127175575-127175575 on Build GRCh38
rs560549895 Chr.10: 127175407-127175407 on Build GRCh38
rs564423956 Chr.10: 127175493-127175493 on Build GRCh38
rs1037576344 Chr.10: 127175387-127175387 on Build GRCh38
rs771416390 Chr.10: 127175431-127175431 on Build GRCh38
rs973403565 Chr.10: 127175545-127175545 on Build GRCh38
rs755171732 Chr.10: 127175413-127175413 on Build GRCh38
rs531616927 Chr.10: 127175494-127175494 on Build GRCh38
rs139302074 Chr.10: 127175427-127175427 on Build GRCh38
rs780666441 Chr.10: 127175564-127175564 on Build GRCh38
rs376061425 Chr.10: 127175533-127175533 on Build GRCh38
rs372852521 Chr.10: 127175499-127175499 on Build GRCh38
rs143223796 Chr.10: 127175447-127175447 on Build GRCh38
rs775162815 Chr.10: 127175442-127175442 on Build GRCh38
rs759630403 Chr.10: 127175467-127175467 on Build GRCh38
rs772328385 Chr.10: 127175369-127175369 on Build GRCh38
rs201089083 Chr.10: 127175459-127175459 on Build GRCh38
rs778283590 Chr.10: 127175372-127175372 on Build GRCh38
rs376501105 Chr.10: 127175561-127175561 on Build GRCh38
rs747492162 Chr.10: 127175375-127175375 on Build GRCh38
rs781623674 Chr.10: 127175510-127175510 on Build GRCh38

View Full Product Details