Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50026207-50026458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNEP1R1
Entrez Gene ID
255919
Gene Name
CTD nuclear envelope phosphatase 1 regulatory subunit 1
Gene Aliases
C16orf69, NEP1-R1, NEP1R1, TMEM188, TMP125
UniGene
Hs.59134
Gene Chromosome Location
Chr.16: 50025206-50037088 on Build GRCh38
Transcripts
NM_001281789.1 NM_153261.5 NR_104042.1 NR_104043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1609375 c.138A>T p.G46G Chr.16: 50026457-50026457 on Build GRCh38
5779947 c.105T>C p.T35T Chr.16: 50026424-50026424 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs541895279 Chr.16: 50026235-50026235 on Build GRCh38
rs569209008 Chr.16: 50026377-50026377 on Build GRCh38
rs765285578 Chr.16: 50026379-50026379 on Build GRCh38
rs750912118 Chr.16: 50026417-50026417 on Build GRCh38
rs17213264 Chr.16: 50026292-50026292 on Build GRCh38
rs552364603 Chr.16: 50026373-50026373 on Build GRCh38
rs773057469 Chr.16: 50026367-50026367 on Build GRCh38
rs376368462 Chr.16: 50026352-50026352 on Build GRCh38
rs565589295 Chr.16: 50026311-50026313 on Build GRCh38
rs1016339662 Chr.16: 50026296-50026297 on Build GRCh38
rs764542639 Chr.16: 50026382-50026382 on Build GRCh38
rs17213250 Chr.16: 50026246-50026246 on Build GRCh38
rs766239146 Chr.16: 50026372-50026372 on Build GRCh38
rs781748007 Chr.16: 50026392-50026392 on Build GRCh38
rs994053073 Chr.16: 50026264-50026264 on Build GRCh38
rs371032588 Chr.16: 50026371-50026371 on Build GRCh38
rs747804577 Chr.16: 50026346-50026346 on Build GRCh38
rs965126552 Chr.16: 50026355-50026355 on Build GRCh38
rs373002381 Chr.16: 50026380-50026380 on Build GRCh38
rs576591148 Chr.16: 50026388-50026388 on Build GRCh38
rs185642108 Chr.16: 50026288-50026288 on Build GRCh38
rs769402287 Chr.16: 50026431-50026431 on Build GRCh38
rs760477957 Chr.16: 50026369-50026369 on Build GRCh38
rs897237516 Chr.16: 50026403-50026403 on Build GRCh38
rs1035331366 Chr.16: 50026240-50026240 on Build GRCh38
rs757697295 Chr.16: 50026387-50026387 on Build GRCh38
rs748020845 Chr.16: 50026384-50026384 on Build GRCh38
rs146610622 Chr.16: 50026276-50026276 on Build GRCh38
rs1044801676 Chr.16: 50026338-50026338 on Build GRCh38
rs905838391 Chr.16: 50026361-50026361 on Build GRCh38
rs79550856 Chr.16: 50026239-50026239 on Build GRCh38

View Full Product Details