Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 161139459-161139708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT4
Entrez Gene ID
56895
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 4
Gene Aliases
1-AGPAT4, LPAAT-delta, dJ473J16.2
UniGene
Hs.353175
Gene Chromosome Location
Chr.6: 161129980-161274108 on Build GRCh38
Transcripts
NM_020133.2 XM_005267053.3 XM_006715514.3 XM_017011059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7182281 c.993G>C p.R331S Chr.6: 161139471-161139471 on Build GRCh38
6314982 c.872C>A p.T291K Chr.6: 161139592-161139592 on Build GRCh38
1442082 c.891G>A p.T297T Chr.6: 161139573-161139573 on Build GRCh38
1244648 c.948G>A p.S316S Chr.6: 161139516-161139516 on Build GRCh38
207487 c.907delC p.R303fs*7 Chr.6: 161139557-161139557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779673784 Chr.6: 161139661-161139661 on Build GRCh38
rs370713321 Chr.6: 161139592-161139592 on Build GRCh38
rs750826126 Chr.6: 161139556-161139557 on Build GRCh38
rs144362957 Chr.6: 161139574-161139574 on Build GRCh38
rs774794072 Chr.6: 161139614-161139614 on Build GRCh38
rs746899044 Chr.6: 161139503-161139503 on Build GRCh38
rs566011116 Chr.6: 161139543-161139543 on Build GRCh38
rs201317021 Chr.6: 161139664-161139664 on Build GRCh38
rs750463380 Chr.6: 161139556-161139556 on Build GRCh38
rs373587636 Chr.6: 161139560-161139560 on Build GRCh38
rs1033789646 Chr.6: 161139654-161139654 on Build GRCh38
rs753948470 Chr.6: 161139638-161139638 on Build GRCh38
rs757160130 Chr.6: 161139568-161139568 on Build GRCh38
rs977803845 Chr.6: 161139507-161139507 on Build GRCh38
rs565134470 Chr.6: 161139591-161139591 on Build GRCh38
rs769371717 Chr.6: 161139663-161139663 on Build GRCh38
rs750308344 Chr.6: 161139657-161139657 on Build GRCh38
rs756140825 Chr.6: 161139557-161139557 on Build GRCh38
rs139702200 Chr.6: 161139505-161139505 on Build GRCh38
rs1047728101 Chr.6: 161139590-161139590 on Build GRCh38
rs535322746 Chr.6: 161139539-161139539 on Build GRCh38
rs767988578 Chr.6: 161139623-161139623 on Build GRCh38
rs758156357 Chr.6: 161139483-161139483 on Build GRCh38
rs777502914 Chr.6: 161139561-161139561 on Build GRCh38
rs777590829 Chr.6: 161139495-161139495 on Build GRCh38
rs756101507 Chr.6: 161139658-161139658 on Build GRCh38
rs114144656 Chr.6: 161139540-161139540 on Build GRCh38
rs780976897 Chr.6: 161139569-161139569 on Build GRCh38
rs201134001 Chr.6: 161139553-161139553 on Build GRCh38
rs775190540 Chr.6: 161139587-161139587 on Build GRCh38
rs762101800 Chr.6: 161139620-161139620 on Build GRCh38
rs929506956 Chr.6: 161139588-161139588 on Build GRCh38
rs767919366 Chr.6: 161139554-161139554 on Build GRCh38
rs140505632 Chr.6: 161139573-161139573 on Build GRCh38
rs542686127 Chr.6: 161139656-161139656 on Build GRCh38
rs180707058 Chr.6: 161139632-161139632 on Build GRCh38
rs1039146831 Chr.6: 161139542-161139542 on Build GRCh38
rs765458550 Chr.6: 161139642-161139642 on Build GRCh38
rs202150883 Chr.6: 161139641-161139641 on Build GRCh38
rs756998971 Chr.6: 161139650-161139650 on Build GRCh38
rs776140497 Chr.6: 161139516-161139516 on Build GRCh38
rs766516442 Chr.6: 161139559-161139559 on Build GRCh38
rs761291733 Chr.6: 161139636-161139636 on Build GRCh38
rs768456360 Chr.6: 161139603-161139603 on Build GRCh38
rs746809281 Chr.6: 161139567-161139567 on Build GRCh38
rs377360821 Chr.6: 161139527-161139528 on Build GRCh38
rs369624936 Chr.6: 161139517-161139517 on Build GRCh38
rs769404968 Chr.6: 161139527-161139527 on Build GRCh38
rs775277076 Chr.6: 161139528-161139528 on Build GRCh38
rs149219965 Chr.6: 161139649-161139649 on Build GRCh38

View Full Product Details