Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 56623389-56623614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM208A
Entrez Gene ID
23272
Gene Name
family with sequence similarity 208 member A
Gene Aliases
C3orf63, RAP140, se89-1
UniGene
Hs.116877
Gene Chromosome Location
Chr.3: 56620132-56683242 on Build GRCh38
Transcripts
NM_001112736.1 NM_015224.3 XM_005264999.1 XM_006713077.2 XM_006713078.2 XM_011533552.2 XM_011533553.2 XM_017006030.1 XM_017006031.1 XM_017006032.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6269245 c.4426C>A p.R1476S Chr.3: 56623441-56623441 on Build GRCh38
1617929 c.3246A>C p.E1082D Chr.3: 56623493-56623493 on Build GRCh38
1617931 c.3235T>A p.S1079T Chr.3: 56623504-56623504 on Build GRCh38
4804902 c.4474C>T p.P1492S Chr.3: 56623393-56623393 on Build GRCh38
3660665 c.3235T>C p.S1079P Chr.3: 56623504-56623504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753555079 Chr.3: 56623553-56623553 on Build GRCh38
rs774785004 Chr.3: 56623490-56623490 on Build GRCh38
rs147511651 Chr.3: 56623441-56623441 on Build GRCh38
rs781503760 Chr.3: 56623440-56623440 on Build GRCh38
rs146566038 Chr.3: 56623419-56623419 on Build GRCh38
rs888579109 Chr.3: 56623501-56623501 on Build GRCh38
rs373715260 Chr.3: 56623477-56623477 on Build GRCh38
rs746740153 Chr.3: 56623433-56623433 on Build GRCh38
rs752672265 Chr.3: 56623416-56623416 on Build GRCh38
rs202079284 Chr.3: 56623467-56623467 on Build GRCh38
rs767856680 Chr.3: 56623551-56623551 on Build GRCh38
rs762028222 Chr.3: 56623497-56623497 on Build GRCh38
rs771186607 Chr.3: 56623439-56623439 on Build GRCh38
rs775693891 Chr.3: 56623449-56623449 on Build GRCh38
rs763541497 Chr.3: 56623475-56623475 on Build GRCh38
rs200344458 Chr.3: 56623584-56623584 on Build GRCh38
rs777645041 Chr.3: 56623421-56623421 on Build GRCh38
rs768896673 Chr.3: 56623582-56623582 on Build GRCh38
rs769007001 Chr.3: 56623482-56623482 on Build GRCh38
rs912871429 Chr.3: 56623495-56623495 on Build GRCh38
rs780396137 Chr.3: 56623545-56623553 on Build GRCh38
rs1048920205 Chr.3: 56623462-56623462 on Build GRCh38
rs770062644 Chr.3: 56623446-56623446 on Build GRCh38
rs747607787 Chr.3: 56623548-56623550 on Build GRCh38

View Full Product Details