Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 152972953-152973226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO5
Entrez Gene ID
26271
Gene Name
F-box protein 5
Gene Aliases
EMI1, FBX5, Fbxo31
UniGene
Hs.520506
Gene Chromosome Location
Chr.6: 152970523-152983605 on Build GRCh38
Transcripts
NM_001142522.1 NM_012177.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6991620 c.901A>G p.R301G Chr.6: 152973054-152973054 on Build GRCh38
3715351 c.835A>T p.T279S Chr.6: 152973120-152973120 on Build GRCh38
4974428 c.862G>A p.D288N Chr.6: 152973093-152973093 on Build GRCh38
1075170 c.909C>T p.T303T Chr.6: 152973046-152973046 on Build GRCh38
1075171 c.878A>G p.Q293R Chr.6: 152973077-152973077 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140496847 Chr.6: 152973088-152973088 on Build GRCh38
rs762767554 Chr.6: 152973098-152973098 on Build GRCh38
rs746715283 Chr.6: 152973060-152973060 on Build GRCh38
rs772568744 Chr.6: 152973055-152973055 on Build GRCh38
rs778522277 Chr.6: 152973026-152973028 on Build GRCh38
rs751673872 Chr.6: 152973117-152973117 on Build GRCh38
rs550217048 Chr.6: 152972995-152972995 on Build GRCh38
rs746227583 Chr.6: 152973029-152973029 on Build GRCh38
rs375664979 Chr.6: 152972986-152972986 on Build GRCh38
rs754411452 Chr.6: 152973183-152973183 on Build GRCh38
rs762190172 Chr.6: 152973130-152973130 on Build GRCh38
rs769426159 Chr.6: 152973085-152973085 on Build GRCh38
rs780244949 Chr.6: 152973173-152973173 on Build GRCh38
rs771646562 Chr.6: 152973053-152973053 on Build GRCh38
rs986932419 Chr.6: 152972981-152972981 on Build GRCh38
rs772222363 Chr.6: 152973006-152973006 on Build GRCh38
rs1046214295 Chr.6: 152973107-152973107 on Build GRCh38
rs529806387 Chr.6: 152973076-152973076 on Build GRCh38
rs770559808 Chr.6: 152973062-152973062 on Build GRCh38
rs548499325 Chr.6: 152973156-152973156 on Build GRCh38
rs758526620 Chr.6: 152972996-152972996 on Build GRCh38
rs936647887 Chr.6: 152973025-152973025 on Build GRCh38
rs764039965 Chr.6: 152972997-152972997 on Build GRCh38
rs764281337 Chr.6: 152973100-152973100 on Build GRCh38
rs767853158 Chr.6: 152973143-152973143 on Build GRCh38
rs984198190 Chr.6: 152973140-152973140 on Build GRCh38
rs568735325 Chr.6: 152973051-152973051 on Build GRCh38
rs537731928 Chr.6: 152973007-152973007 on Build GRCh38
rs776478627 Chr.6: 152973068-152973068 on Build GRCh38
rs532089118 Chr.6: 152972993-152972993 on Build GRCh38
rs780619610 Chr.6: 152973046-152973046 on Build GRCh38
rs781726545 Chr.6: 152973027-152973027 on Build GRCh38
rs954195889 Chr.6: 152973109-152973109 on Build GRCh38
rs372476270 Chr.6: 152973031-152973031 on Build GRCh38
rs759331223 Chr.6: 152973077-152973077 on Build GRCh38
rs976579297 Chr.6: 152973047-152973047 on Build GRCh38
rs771000854 Chr.6: 152973042-152973042 on Build GRCh38
rs370586059 Chr.6: 152973015-152973015 on Build GRCh38
rs942757381 Chr.6: 152972987-152972987 on Build GRCh38

View Full Product Details