Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149349577-149349824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRPEL2
Entrez Gene ID
134266
Gene Name
GrpE like 2, mitochondrial
Gene Aliases
Mt-GrpE#2
UniGene
Hs.511816
Gene Chromosome Location
Chr.5: 149345414-149354583 on Build GRCh38
Transcripts
NM_152407.3 NR_132366.1

Gene Symbol
GRPEL2-AS1
Entrez Gene ID
106144529
Gene Name
GRPEL2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.607162
Gene Chromosome Location
Chr.5: 149348116-149357642 on Build GRCh38
Transcripts
NM_152407.3 NR_132366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5356404 c.281C>T p.T94I Chr.5: 149349703-149349703 on Build GRCh38
1209085 c.286A>G p.R96G Chr.5: 149349708-149349708 on Build GRCh38
5551412 c.291T>C p.C97C Chr.5: 149349713-149349713 on Build GRCh38
5402662 c.240C>T p.Y80Y Chr.5: 149349662-149349662 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768453879 Chr.5: 149349760-149349760 on Build GRCh38
rs200267116 Chr.5: 149349761-149349761 on Build GRCh38
rs771977405 Chr.5: 149349747-149349747 on Build GRCh38
rs151163426 Chr.5: 149349665-149349665 on Build GRCh38
rs753905171 Chr.5: 149349609-149349609 on Build GRCh38
rs772439459 Chr.5: 149349777-149349777 on Build GRCh38
rs751617949 Chr.5: 149349786-149349786 on Build GRCh38
rs770758108 Chr.5: 149349742-149349742 on Build GRCh38
rs750943165 Chr.5: 149349699-149349699 on Build GRCh38
rs146069925 Chr.5: 149349722-149349722 on Build GRCh38
rs371769490 Chr.5: 149349745-149349745 on Build GRCh38
rs755173203 Chr.5: 149349732-149349732 on Build GRCh38
rs778663026 Chr.5: 149349744-149349744 on Build GRCh38
rs561635422 Chr.5: 149349792-149349792 on Build GRCh38
rs765666161 Chr.5: 149349676-149349676 on Build GRCh38
rs553077407 Chr.5: 149349609-149349609 on Build GRCh38
rs755086886 Chr.5: 149349789-149349789 on Build GRCh38
rs759678955 Chr.5: 149349692-149349694 on Build GRCh38
rs960335891 Chr.5: 149349787-149349787 on Build GRCh38
rs748334239 Chr.5: 149349740-149349740 on Build GRCh38
rs374175991 Chr.5: 149349675-149349675 on Build GRCh38
rs368140281 Chr.5: 149349607-149349607 on Build GRCh38
rs140331718 Chr.5: 149349667-149349667 on Build GRCh38
rs142275296 Chr.5: 149349700-149349700 on Build GRCh38
rs758671882 Chr.5: 149349765-149349766 on Build GRCh38
rs777957866 Chr.5: 149349786-149349794 on Build GRCh38
rs765756206 Chr.5: 149349768-149349768 on Build GRCh38
rs770882934 Chr.5: 149349610-149349610 on Build GRCh38
rs778017400 Chr.5: 149349780-149349780 on Build GRCh38
rs773623727 Chr.5: 149349775-149349775 on Build GRCh38
rs150347105 Chr.5: 149349671-149349671 on Build GRCh38
rs768922633 Chr.5: 149349606-149349606 on Build GRCh38
rs780067271 Chr.5: 149349709-149349709 on Build GRCh38
rs781347032 Chr.5: 149349739-149349739 on Build GRCh38
rs774310453 Chr.5: 149349617-149349617 on Build GRCh38
rs766853712 Chr.5: 149349783-149349783 on Build GRCh38
rs200636189 Chr.5: 149349668-149349668 on Build GRCh38
rs761535980 Chr.5: 149349762-149349762 on Build GRCh38
rs369193538 Chr.5: 149349753-149349753 on Build GRCh38
rs775060667 Chr.5: 149349663-149349663 on Build GRCh38
rs187777262 Chr.5: 149349661-149349661 on Build GRCh38
rs752860375 Chr.5: 149349756-149349756 on Build GRCh38
rs187464359 Chr.5: 149349778-149349778 on Build GRCh38
rs767558027 Chr.5: 149349798-149349798 on Build GRCh38
rs760182229 Chr.5: 149349755-149349755 on Build GRCh38
rs765305568 Chr.5: 149349738-149349739 on Build GRCh38
rs138111749 Chr.5: 149349672-149349672 on Build GRCh38
rs759491517 Chr.5: 149349634-149349634 on Build GRCh38

View Full Product Details