Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150948703-150948976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH2
Entrez Gene ID
3757
Gene Name
potassium voltage-gated channel subfamily H member 2
Gene Aliases
ERG-1, ERG1, H-ERG, HERG, HERG1, Kv11.1, LQT2, SQT1
UniGene
Hs.647099
Gene Chromosome Location
Chr.7: 150944956-150978314 on Build GRCh38
Transcripts
NM_000238.3 NM_172057.2 XM_011516185.2 XM_011516186.2 XM_017012195.1 XM_017012196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4694321 c.1567C>T p.R523* Chr.7: 150948861-150948861 on Build GRCh38
4538565 c.2569G>A p.E857K Chr.7: 150948879-150948879 on Build GRCh38
6690859 c.1524C>A p.F508L Chr.7: 150948904-150948904 on Build GRCh38
4907346 c.1531C>G p.H511D Chr.7: 150948897-150948897 on Build GRCh38
6202786 c.1450_1486del37 p.A484fs*32 Chr.7: 150948942-150948978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774390249 Chr.7: 150948827-150948827 on Build GRCh38
rs199473006 Chr.7: 150948912-150948912 on Build GRCh38
rs543184784 Chr.7: 150948901-150948901 on Build GRCh38
rs754565340 Chr.7: 150948918-150948918 on Build GRCh38
rs111531831 Chr.7: 150948913-150948913 on Build GRCh38
rs939435672 Chr.7: 150948860-150948860 on Build GRCh38
rs758040682 Chr.7: 150948947-150948947 on Build GRCh38
rs139295242 Chr.7: 150948940-150948940 on Build GRCh38
rs529469304 Chr.7: 150948802-150948802 on Build GRCh38
rs762794122 Chr.7: 150948745-150948745 on Build GRCh38
rs746785851 Chr.7: 150948808-150948808 on Build GRCh38
rs121912513 Chr.7: 150948866-150948866 on Build GRCh38
rs948648427 Chr.7: 150948753-150948753 on Build GRCh38
rs753483535 Chr.7: 150948910-150948910 on Build GRCh38
rs752253646 Chr.7: 150948930-150948930 on Build GRCh38
rs371624920 Chr.7: 150948821-150948821 on Build GRCh38
rs199473539 Chr.7: 150948875-150948875 on Build GRCh38
rs199473005 Chr.7: 150948939-150948939 on Build GRCh38
rs1036885088 Chr.7: 150948900-150948900 on Build GRCh38
rs773724817 Chr.7: 150948861-150948861 on Build GRCh38
rs200451965 Chr.7: 150948845-150948845 on Build GRCh38
rs761038306 Chr.7: 150948876-150948876 on Build GRCh38
rs199473003 Chr.7: 150948945-150948945 on Build GRCh38
rs794728395 Chr.7: 150948944-150948944 on Build GRCh38
rs866263680 Chr.7: 150948907-150948907 on Build GRCh38
rs906562788 Chr.7: 150948853-150948853 on Build GRCh38
rs794728393 Chr.7: 150948954-150948954 on Build GRCh38
rs577567071 Chr.7: 150948752-150948752 on Build GRCh38
rs1036535206 Chr.7: 150948859-150948859 on Build GRCh38
rs794728396 Chr.7: 150948872-150948872 on Build GRCh38
rs768003897 Chr.7: 150948846-150948846 on Build GRCh38
rs199473004 Chr.7: 150948938-150948938 on Build GRCh38
rs563605594 Chr.7: 150948776-150948776 on Build GRCh38
rs897540177 Chr.7: 150948886-150948886 on Build GRCh38
rs199473007 Chr.7: 150948867-150948867 on Build GRCh38
rs895454791 Chr.7: 150948747-150948747 on Build GRCh38
rs770644364 Chr.7: 150948813-150948813 on Build GRCh38
rs199473008 Chr.7: 150948857-150948857 on Build GRCh38
rs559217960 Chr.7: 150948920-150948920 on Build GRCh38
rs775713207 Chr.7: 150948819-150948819 on Build GRCh38
rs543714814 Chr.7: 150948757-150948757 on Build GRCh38
rs768730376 Chr.7: 150948822-150948822 on Build GRCh38

View Full Product Details