Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73385776-73386001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALMS1
Entrez Gene ID
7840
Gene Name
ALMS1, centrosome and basal body associated protein
Gene Aliases
ALSS
UniGene
Hs.184720
Gene Chromosome Location
Chr.2: 73385758-73609919 on Build GRCh38
Transcripts
NM_015120.4

SNP Detail

SNP ID Chromosome Location
rs548574576 Chr.2: 73385852-73385852 on Build GRCh38
rs1053425100 Chr.2: 73385875-73385875 on Build GRCh38
rs906421049 Chr.2: 73385920-73385920 on Build GRCh38
rs754016018 Chr.2: 73385831-73385831 on Build GRCh38
rs1006328829 Chr.2: 73385935-73385936 on Build GRCh38
rs767882166 Chr.2: 73385925-73385926 on Build GRCh38
rs1045405319 Chr.2: 73385896-73385896 on Build GRCh38
rs768547463 Chr.2: 73385828-73385829 on Build GRCh38
rs777844728 Chr.2: 73385932-73385933 on Build GRCh38
rs780250336 Chr.2: 73385830-73385830 on Build GRCh38
rs70965731 Chr.2: 73385940-73385942 on Build GRCh38
rs886056296 Chr.2: 73385970-73385971 on Build GRCh38
rs905301935 Chr.2: 73385929-73385930 on Build GRCh38
rs878854999 Chr.2: 73385925-73385942 on Build GRCh38
rs1054040147 Chr.2: 73385937-73385937 on Build GRCh38
rs1016268902 Chr.2: 73385826-73385826 on Build GRCh38
rs774968380 Chr.2: 73385954-73385954 on Build GRCh38
rs61156725 Chr.2: 73385937-73385942 on Build GRCh38
rs373614102 Chr.2: 73385889-73385889 on Build GRCh38
rs767731236 Chr.2: 73385982-73385982 on Build GRCh38
rs955371491 Chr.2: 73385971-73385971 on Build GRCh38
rs781075077 Chr.2: 73385874-73385874 on Build GRCh38
rs754175473 Chr.2: 73385938-73385943 on Build GRCh38
rs1025593035 Chr.2: 73385956-73385956 on Build GRCh38
rs543687863 Chr.2: 73385821-73385821 on Build GRCh38
rs797045229 Chr.2: 73385945-73385953 on Build GRCh38
rs755530002 Chr.2: 73385919-73385923 on Build GRCh38
rs780977235 Chr.2: 73385959-73385964 on Build GRCh38
rs955065937 Chr.2: 73385839-73385839 on Build GRCh38
rs935547269 Chr.2: 73385849-73385849 on Build GRCh38
rs774784126 Chr.2: 73385977-73385977 on Build GRCh38
rs193922695 Chr.2: 73385939-73385940 on Build GRCh38
rs866503570 Chr.2: 73385925-73385925 on Build GRCh38
rs768161233 Chr.2: 73385962-73385962 on Build GRCh38
rs576062106 Chr.2: 73385807-73385807 on Build GRCh38
rs756956183 Chr.2: 73385868-73385868 on Build GRCh38
rs886056294 Chr.2: 73385830-73385830 on Build GRCh38
rs746896173 Chr.2: 73385958-73385959 on Build GRCh38
rs756301830 Chr.2: 73385931-73385932 on Build GRCh38
rs746980000 Chr.2: 73385829-73385831 on Build GRCh38
rs1044954486 Chr.2: 73385797-73385797 on Build GRCh38
rs878853007 Chr.2: 73385907-73385909 on Build GRCh38
rs548057920 Chr.2: 73385828-73385828 on Build GRCh38
rs199611703 Chr.2: 73385902-73385902 on Build GRCh38
rs764473588 Chr.2: 73385824-73385824 on Build GRCh38
rs779179743 Chr.2: 73385943-73385944 on Build GRCh38
rs13009609 Chr.2: 73385961-73385961 on Build GRCh38
rs914898490 Chr.2: 73385876-73385876 on Build GRCh38
rs753006733 Chr.2: 73385928-73385928 on Build GRCh38
rs764804760 Chr.2: 73385903-73385904 on Build GRCh38
rs879255406 Chr.2: 73385937-73385939 on Build GRCh38
rs752032517 Chr.2: 73385916-73385916 on Build GRCh38
rs762403314 Chr.2: 73385978-73385978 on Build GRCh38
rs939208094 Chr.2: 73385923-73385923 on Build GRCh38
rs1004961829 Chr.2: 73385931-73385931 on Build GRCh38
rs746944958 Chr.2: 73385853-73385853 on Build GRCh38
rs778948871 Chr.2: 73385909-73385910 on Build GRCh38
rs756389923 Chr.2: 73385829-73385829 on Build GRCh38
rs200709146 Chr.2: 73385903-73385903 on Build GRCh38
rs772724963 Chr.2: 73385904-73385906 on Build GRCh38
rs1016245881 Chr.2: 73385948-73385948 on Build GRCh38
rs1003354672 Chr.2: 73385802-73385802 on Build GRCh38
rs918382598 Chr.2: 73385824-73385826 on Build GRCh38
rs13009043 Chr.2: 73385943-73385943 on Build GRCh38
rs864622738 Chr.2: 73385934-73385942 on Build GRCh38
rs886056293 Chr.2: 73385805-73385805 on Build GRCh38
rs878855000 Chr.2: 73385928-73385942 on Build GRCh38
rs1023762464 Chr.2: 73385805-73385805 on Build GRCh38
rs1035787897 Chr.2: 73385811-73385811 on Build GRCh38
rs530120421 Chr.2: 73385848-73385848 on Build GRCh38
rs762436798 Chr.2: 73385904-73385915 on Build GRCh38
rs987571770 Chr.2: 73385840-73385840 on Build GRCh38
rs539250179 Chr.2: 73385824-73385824 on Build GRCh38
rs886056295 Chr.2: 73385842-73385844 on Build GRCh38
rs769538172 Chr.2: 73385893-73385893 on Build GRCh38
rs748863552 Chr.2: 73385955-73385955 on Build GRCh38
rs570180164 Chr.2: 73385877-73385877 on Build GRCh38
rs13009604 Chr.2: 73385946-73385946 on Build GRCh38
rs183407241 Chr.2: 73385928-73385928 on Build GRCh38
rs868088238 Chr.2: 73385816-73385816 on Build GRCh38
rs554379302 Chr.2: 73385842-73385842 on Build GRCh38
rs386390463 Chr.2: 73385922-73385923 on Build GRCh38
rs55889738 Chr.2: 73385942-73385943 on Build GRCh38
rs973057702 Chr.2: 73385956-73385958 on Build GRCh38
rs962597080 Chr.2: 73385823-73385824 on Build GRCh38
rs969372310 Chr.2: 73385815-73385816 on Build GRCh38
rs906360921 Chr.2: 73385801-73385801 on Build GRCh38

View Full Product Details