Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157738371-157738644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1
Entrez Gene ID
90
Gene Name
activin A receptor type 1
Gene Aliases
ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI
UniGene
Hs.470316
Gene Chromosome Location
Chr.2: 157736446-157875880 on Build GRCh38
Transcripts
NM_001105.4 NM_001111067.2 XM_005246939.3 XM_005246940.3 XM_006712825.3 XM_011512106.2 XM_011512107.2 XM_011512108.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3961332 c.1361G>T p.R454M Chr.2: 157738474-157738474 on Build GRCh38
4976266 c.1269A>G p.I423M Chr.2: 157738566-157738566 on Build GRCh38
5476232 c.1368C>T p.N456N Chr.2: 157738467-157738467 on Build GRCh38
3299840 c.1318A>G p.S440G Chr.2: 157738517-157738517 on Build GRCh38
4448877 c.1395+1G>A p.? Chr.2: 157738439-157738439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568204852 Chr.2: 157738549-157738549 on Build GRCh38
rs781232217 Chr.2: 157738466-157738466 on Build GRCh38
rs368919349 Chr.2: 157738598-157738598 on Build GRCh38
rs1011142075 Chr.2: 157738467-157738467 on Build GRCh38
rs143068325 Chr.2: 157738545-157738545 on Build GRCh38
rs762094464 Chr.2: 157738435-157738435 on Build GRCh38
rs756877324 Chr.2: 157738494-157738494 on Build GRCh38
rs777678249 Chr.2: 157738485-157738485 on Build GRCh38
rs903745092 Chr.2: 157738408-157738408 on Build GRCh38
rs763076822 Chr.2: 157738592-157738592 on Build GRCh38
rs554787560 Chr.2: 157738597-157738597 on Build GRCh38
rs761950656 Chr.2: 157738550-157738550 on Build GRCh38
rs375101352 Chr.2: 157738525-157738525 on Build GRCh38
rs765430692 Chr.2: 157738551-157738551 on Build GRCh38
rs763994830 Chr.2: 157738617-157738617 on Build GRCh38
rs372382136 Chr.2: 157738611-157738611 on Build GRCh38
rs750457181 Chr.2: 157738441-157738441 on Build GRCh38
rs373855918 Chr.2: 157738440-157738440 on Build GRCh38
rs148153887 Chr.2: 157738475-157738475 on Build GRCh38
rs886042941 Chr.2: 157738443-157738443 on Build GRCh38
rs775864239 Chr.2: 157738414-157738414 on Build GRCh38
rs773193666 Chr.2: 157738580-157738580 on Build GRCh38
rs754871846 Chr.2: 157738458-157738458 on Build GRCh38
rs760982754 Chr.2: 157738422-157738422 on Build GRCh38
rs370008141 Chr.2: 157738425-157738425 on Build GRCh38
rs984929600 Chr.2: 157738590-157738590 on Build GRCh38
rs201131823 Chr.2: 157738539-157738539 on Build GRCh38
rs745343863 Chr.2: 157738517-157738517 on Build GRCh38
rs764423594 Chr.2: 157738424-157738424 on Build GRCh38
rs756115532 Chr.2: 157738614-157738614 on Build GRCh38
rs773595846 Chr.2: 157738426-157738437 on Build GRCh38
rs769079595 Chr.2: 157738540-157738540 on Build GRCh38
rs1026045983 Chr.2: 157738537-157738537 on Build GRCh38
rs776029462 Chr.2: 157738538-157738538 on Build GRCh38
rs778291319 Chr.2: 157738500-157738500 on Build GRCh38
rs778133291 Chr.2: 157738433-157738433 on Build GRCh38
rs202095604 Chr.2: 157738535-157738535 on Build GRCh38
rs527887447 Chr.2: 157738491-157738491 on Build GRCh38
rs756022457 Chr.2: 157738480-157738480 on Build GRCh38

View Full Product Details