Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 70262662-70262935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13C
Entrez Gene ID
81573
Gene Name
ankyrin repeat domain 13C
Gene Aliases
dJ677H15.3
UniGene
Hs.744989
Gene Chromosome Location
Chr.1: 70259002-70354977 on Build GRCh38
Transcripts
NM_030816.4 XM_005271234.3 XM_005271235.3 XM_006710929.3 XM_017002413.1 XM_017002414.1 XM_017002415.1 XM_017002416.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6460678 c.1555G>A p.E519K Chr.1: 70262788-70262788 on Build GRCh38
6600385 c.1546C>T p.R516* Chr.1: 70262797-70262797 on Build GRCh38
414797 c.1561G>A p.D521N Chr.1: 70262782-70262782 on Build GRCh38
6600383 c.1609C>T p.R537C Chr.1: 70262734-70262734 on Build GRCh38
5735015 c.1516A>G p.I506V Chr.1: 70262827-70262827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749384591 Chr.1: 70262774-70262774 on Build GRCh38
rs747184413 Chr.1: 70262855-70262855 on Build GRCh38
rs762387724 Chr.1: 70262712-70262712 on Build GRCh38
rs545714875 Chr.1: 70262831-70262831 on Build GRCh38
rs774034864 Chr.1: 70262894-70262894 on Build GRCh38
rs770670707 Chr.1: 70262796-70262796 on Build GRCh38
rs776480050 Chr.1: 70262825-70262825 on Build GRCh38
rs74373835 Chr.1: 70262826-70262826 on Build GRCh38
rs767896416 Chr.1: 70262744-70262744 on Build GRCh38
rs866800802 Chr.1: 70262803-70262803 on Build GRCh38
rs144692308 Chr.1: 70262696-70262696 on Build GRCh38
rs755071399 Chr.1: 70262789-70262789 on Build GRCh38
rs377639211 Chr.1: 70262861-70262861 on Build GRCh38
rs1018194646 Chr.1: 70262739-70262739 on Build GRCh38
rs373023045 Chr.1: 70262697-70262697 on Build GRCh38
rs201846288 Chr.1: 70262716-70262716 on Build GRCh38
rs190340576 Chr.1: 70262864-70262864 on Build GRCh38
rs1018716021 Chr.1: 70262709-70262709 on Build GRCh38
rs926185563 Chr.1: 70262790-70262790 on Build GRCh38
rs779302047 Chr.1: 70262791-70262791 on Build GRCh38
rs751170969 Chr.1: 70262740-70262740 on Build GRCh38
rs756635238 Chr.1: 70262748-70262748 on Build GRCh38
rs763544193 Chr.1: 70262876-70262876 on Build GRCh38
rs528399681 Chr.1: 70262866-70262866 on Build GRCh38
rs114406540 Chr.1: 70262874-70262874 on Build GRCh38
rs140092352 Chr.1: 70262750-70262750 on Build GRCh38
rs752185926 Chr.1: 70262711-70262711 on Build GRCh38
rs757670074 Chr.1: 70262741-70262741 on Build GRCh38
rs776272602 Chr.1: 70262715-70262718 on Build GRCh38
rs186623984 Chr.1: 70262792-70262792 on Build GRCh38
rs979076017 Chr.1: 70262837-70262837 on Build GRCh38
rs974948181 Chr.1: 70262736-70262736 on Build GRCh38

View Full Product Details