Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8473543-8473812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPM
Entrez Gene ID
4670
Gene Name
heterogeneous nuclear ribonucleoprotein M
Gene Aliases
CEAR, HNRNPM4, HNRPM, HNRPM4, HTGR1, NAGR1, hnRNP M
UniGene
Hs.465808
Gene Chromosome Location
Chr.19: 8444575-8489118 on Build GRCh38
Transcripts
NM_001297418.1 NM_005968.4 NM_031203.3 XM_005272478.2 XM_005272479.2 XM_005272480.2 XM_005272481.1 XM_005272483.2 XM_017026823.1 XM_017026824.1 XM_017026825.1 XM_017026826.1 XM_017026827.1 XM_017026828.1 XM_017026829.1 XM_017026830.1 XM_017026831.1 XM_017026832.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5728230 c.1009G>A p.E337K Chr.19: 8473675-8473675 on Build GRCh38
3541613 c.1025G>T p.G342V Chr.19: 8473691-8473691 on Build GRCh38
6681368 c.1030A>C p.N344H Chr.19: 8473696-8473696 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753250759 Chr.19: 8473714-8473715 on Build GRCh38
rs766655475 Chr.19: 8473683-8473683 on Build GRCh38
rs767710669 Chr.19: 8473686-8473686 on Build GRCh38
rs913156590 Chr.19: 8473594-8473594 on Build GRCh38
rs948474408 Chr.19: 8473581-8473581 on Build GRCh38
rs77353381 Chr.19: 8473614-8473614 on Build GRCh38
rs770537227 Chr.19: 8473637-8473637 on Build GRCh38
rs777249768 Chr.19: 8473628-8473628 on Build GRCh38
rs201294620 Chr.19: 8473629-8473629 on Build GRCh38
rs76928964 Chr.19: 8473781-8473781 on Build GRCh38
rs765472975 Chr.19: 8473661-8473661 on Build GRCh38
rs374774628 Chr.19: 8473649-8473649 on Build GRCh38
rs560085376 Chr.19: 8473592-8473592 on Build GRCh38
rs78449787 Chr.19: 8473783-8473783 on Build GRCh38
rs775500833 Chr.19: 8473652-8473652 on Build GRCh38
rs777302903 Chr.19: 8473711-8473711 on Build GRCh38
rs746994177 Chr.19: 8473751-8473751 on Build GRCh38
rs79832765 Chr.19: 8473766-8473766 on Build GRCh38
rs891720723 Chr.19: 8473610-8473610 on Build GRCh38
rs988666804 Chr.19: 8473593-8473593 on Build GRCh38
rs771967222 Chr.19: 8473743-8473743 on Build GRCh38
rs907839003 Chr.19: 8473650-8473651 on Build GRCh38
rs775136806 Chr.19: 8473750-8473750 on Build GRCh38
rs906911743 Chr.19: 8473586-8473586 on Build GRCh38
rs537262489 Chr.19: 8473658-8473658 on Build GRCh38
rs895290972 Chr.19: 8473576-8473576 on Build GRCh38
rs964544438 Chr.19: 8473615-8473615 on Build GRCh38
rs1044089662 Chr.19: 8473582-8473582 on Build GRCh38
rs921698113 Chr.19: 8473606-8473606 on Build GRCh38
rs760305459 Chr.19: 8473649-8473654 on Build GRCh38
rs755386909 Chr.19: 8473685-8473685 on Build GRCh38
rs750371555 Chr.19: 8473611-8473611 on Build GRCh38
rs763307483 Chr.19: 8473667-8473667 on Build GRCh38
rs773831737 Chr.19: 8473644-8473644 on Build GRCh38
rs751687668 Chr.19: 8473684-8473684 on Build GRCh38
rs974859897 Chr.19: 8473622-8473622 on Build GRCh38
rs745698568 Chr.19: 8473645-8473645 on Build GRCh38
rs748822679 Chr.19: 8473712-8473712 on Build GRCh38
rs1002476855 Chr.19: 8473587-8473587 on Build GRCh38
rs778194394 Chr.19: 8473763-8473766 on Build GRCh38
rs773337934 Chr.19: 8473595-8473595 on Build GRCh38
rs976185263 Chr.19: 8473601-8473601 on Build GRCh38
rs1044164247 Chr.19: 8473736-8473736 on Build GRCh38
rs756589175 Chr.19: 8473707-8473707 on Build GRCh38
rs140407642 Chr.19: 8473704-8473704 on Build GRCh38
rs896748417 Chr.19: 8473702-8473702 on Build GRCh38
rs760683368 Chr.19: 8473655-8473655 on Build GRCh38
rs756707655 Chr.19: 8473728-8473728 on Build GRCh38
rs778639553 Chr.19: 8473734-8473734 on Build GRCh38

View Full Product Details