Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 52907753-52908026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHDC2
Entrez Gene ID
55268
Gene Name
enoyl-CoA hydratase domain containing 2
Gene Aliases
-
UniGene
Hs.476319
Gene Chromosome Location
Chr.1: 52895910-52927212 on Build GRCh38
Transcripts
NM_001198961.1 NM_001198962.1 NM_001319958.1 NM_018281.3 XM_011541709.2 XM_011541710.2 XM_011541711.2 XM_011541712.2 XM_011541713.2 XM_011541715.2 XM_011541719.2 XM_011541720.2 XM_011541721.2 XM_011541722.2 XM_011541723.2 XM_011541724.2 XM_011541725.2 XM_011541726.2 XM_011541727.2 XM_017001638.1 XM_017001639.1 XM_017001640.1 XM_017001641.1 XM_017001642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4823146 c.354G>A p.M118I Chr.1: 52907878-52907878 on Build GRCh38
1492129 c.362T>C p.I121T Chr.1: 52907870-52907870 on Build GRCh38
3490692 c.333C>T p.V111V Chr.1: 52907899-52907899 on Build GRCh38
4823148 c.216G>A p.M72I Chr.1: 52907878-52907878 on Build GRCh38
4823147 c.354G>A p.M118I Chr.1: 52907878-52907878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111305550 Chr.1: 52907867-52907867 on Build GRCh38
rs905187844 Chr.1: 52907785-52907785 on Build GRCh38
rs773242791 Chr.1: 52907894-52907894 on Build GRCh38
rs368512350 Chr.1: 52907895-52907895 on Build GRCh38
rs141163827 Chr.1: 52907929-52907929 on Build GRCh38
rs766878998 Chr.1: 52907833-52907833 on Build GRCh38
rs751067095 Chr.1: 52907911-52907911 on Build GRCh38
rs772335252 Chr.1: 52907890-52907890 on Build GRCh38
rs990386177 Chr.1: 52907814-52907814 on Build GRCh38
rs770085379 Chr.1: 52907906-52907907 on Build GRCh38
rs770326097 Chr.1: 52908004-52908004 on Build GRCh38
rs112246621 Chr.1: 52907866-52907866 on Build GRCh38
rs763560505 Chr.1: 52907909-52907909 on Build GRCh38
rs199548521 Chr.1: 52907900-52907900 on Build GRCh38
rs369927995 Chr.1: 52907977-52907977 on Build GRCh38
rs1024088809 Chr.1: 52907784-52907784 on Build GRCh38
rs773615879 Chr.1: 52907828-52907828 on Build GRCh38
rs777153706 Chr.1: 52907903-52907903 on Build GRCh38
rs747190912 Chr.1: 52907991-52907991 on Build GRCh38
rs879238267 Chr.1: 52907852-52907852 on Build GRCh38
rs928553442 Chr.1: 52907856-52907856 on Build GRCh38
rs748388030 Chr.1: 52907841-52907844 on Build GRCh38
rs780150306 Chr.1: 52907869-52907869 on Build GRCh38
rs754286175 Chr.1: 52907836-52907836 on Build GRCh38
rs1031683190 Chr.1: 52907781-52907782 on Build GRCh38
rs774472106 Chr.1: 52907826-52907826 on Build GRCh38
rs749202048 Chr.1: 52907871-52907871 on Build GRCh38
rs755038223 Chr.1: 52907827-52907828 on Build GRCh38
rs776957819 Chr.1: 52908003-52908003 on Build GRCh38
rs779967734 Chr.1: 52907940-52907940 on Build GRCh38
rs761238474 Chr.1: 52907830-52907830 on Build GRCh38
rs368400112 Chr.1: 52907988-52907988 on Build GRCh38
rs867620188 Chr.1: 52907947-52907947 on Build GRCh38
rs771322600 Chr.1: 52907999-52907999 on Build GRCh38
rs755991925 Chr.1: 52907937-52907937 on Build GRCh38
rs778930934 Chr.1: 52907966-52907966 on Build GRCh38
rs548248043 Chr.1: 52907827-52907827 on Build GRCh38
rs755901852 Chr.1: 52907868-52907868 on Build GRCh38
rs566505704 Chr.1: 52907815-52907815 on Build GRCh38
rs113121140 Chr.1: 52907820-52907820 on Build GRCh38
rs981404355 Chr.1: 52907837-52907837 on Build GRCh38
rs1003891859 Chr.1: 52907808-52907808 on Build GRCh38
rs200830326 Chr.1: 52907910-52907910 on Build GRCh38
rs753877882 Chr.1: 52907960-52907960 on Build GRCh38
rs962489552 Chr.1: 52907915-52907915 on Build GRCh38
rs957453882 Chr.1: 52907813-52907813 on Build GRCh38
rs372381507 Chr.1: 52907905-52907905 on Build GRCh38
rs751496619 Chr.1: 52907823-52907824 on Build GRCh38
rs757925344 Chr.1: 52907841-52907841 on Build GRCh38
rs761435615 Chr.1: 52907918-52907918 on Build GRCh38
rs768901238 Chr.1: 52907886-52907886 on Build GRCh38
rs781384773 Chr.1: 52907839-52907841 on Build GRCh38
rs200045876 Chr.1: 52907936-52907936 on Build GRCh38
rs373840820 Chr.1: 52907850-52907850 on Build GRCh38
rs745668272 Chr.1: 52907865-52907865 on Build GRCh38
rs568116482 Chr.1: 52907844-52907844 on Build GRCh38
rs780903338 Chr.1: 52907862-52907862 on Build GRCh38
rs975189335 Chr.1: 52907923-52907923 on Build GRCh38
rs17854314 Chr.1: 52907877-52907877 on Build GRCh38
rs200827816 Chr.1: 52907889-52907889 on Build GRCh38
rs552471016 Chr.1: 52907965-52907965 on Build GRCh38
rs533977217 Chr.1: 52907847-52907847 on Build GRCh38
rs111794011 Chr.1: 52907888-52907888 on Build GRCh38
rs374560400 Chr.1: 52907846-52907846 on Build GRCh38

View Full Product Details