Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 109739328-109739599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSTM3
Entrez Gene ID
2947
Gene Name
glutathione S-transferase mu 3
Gene Aliases
GST5, GSTB, GSTM3-3, GTM3
UniGene
Hs.2006
Gene Chromosome Location
Chr.1: 109733932-109741038 on Build GRCh38
Transcripts
NM_000849.4 NR_024537.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3731993 c.140G>A p.R47Q Chr.1: 109739478-109739478 on Build GRCh38
4689127 c.185C>A p.P62H Chr.1: 109739433-109739433 on Build GRCh38
5843010 c.178delG p.D60fs*45 Chr.1: 109739440-109739440 on Build GRCh38
7102853 c.136G>A p.D46N Chr.1: 109739482-109739482 on Build GRCh38
1332505 c.139C>T p.R47* Chr.1: 109739479-109739479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768554660 Chr.1: 109739380-109739380 on Build GRCh38
rs772169956 Chr.1: 109739401-109739401 on Build GRCh38
rs778809423 Chr.1: 109739451-109739451 on Build GRCh38
rs537537161 Chr.1: 109739385-109739385 on Build GRCh38
rs766497422 Chr.1: 109739432-109739432 on Build GRCh38
rs774651518 Chr.1: 109739499-109739499 on Build GRCh38
rs770536480 Chr.1: 109739477-109739477 on Build GRCh38
rs893104087 Chr.1: 109739444-109739444 on Build GRCh38
rs763690615 Chr.1: 109739357-109739357 on Build GRCh38
rs748032746 Chr.1: 109739386-109739386 on Build GRCh38
rs777894649 Chr.1: 109739471-109739471 on Build GRCh38
rs770462521 Chr.1: 109739405-109739405 on Build GRCh38
rs778587657 Chr.1: 109739382-109739382 on Build GRCh38
rs756257189 Chr.1: 109739426-109739426 on Build GRCh38
rs200315626 Chr.1: 109739504-109739504 on Build GRCh38
rs375148438 Chr.1: 109739377-109739377 on Build GRCh38
rs755158844 Chr.1: 109739450-109739450 on Build GRCh38
rs769364875 Chr.1: 109739493-109739493 on Build GRCh38
rs567631050 Chr.1: 109739406-109739406 on Build GRCh38
rs553483585 Chr.1: 109739527-109739527 on Build GRCh38
rs767981531 Chr.1: 109739521-109739521 on Build GRCh38
rs745461176 Chr.1: 109739485-109739485 on Build GRCh38
rs202087892 Chr.1: 109739484-109739484 on Build GRCh38
rs774700431 Chr.1: 109739414-109739414 on Build GRCh38
rs754033213 Chr.1: 109739435-109739435 on Build GRCh38
rs754954461 Chr.1: 109739531-109739543 on Build GRCh38
rs1005710910 Chr.1: 109739529-109739529 on Build GRCh38
rs368764245 Chr.1: 109739404-109739404 on Build GRCh38
rs747977133 Chr.1: 109739453-109739453 on Build GRCh38
rs11555177 Chr.1: 109739476-109739476 on Build GRCh38
rs78032372 Chr.1: 109739496-109739496 on Build GRCh38
rs967595518 Chr.1: 109739501-109739502 on Build GRCh38
rs751435829 Chr.1: 109739372-109739372 on Build GRCh38
rs1019752693 Chr.1: 109739537-109739537 on Build GRCh38
rs932922040 Chr.1: 109739389-109739389 on Build GRCh38
rs758384444 Chr.1: 109739470-109739470 on Build GRCh38
rs189263054 Chr.1: 109739540-109739540 on Build GRCh38
rs764458914 Chr.1: 109739409-109739409 on Build GRCh38
rs372573919 Chr.1: 109739421-109739421 on Build GRCh38
rs551306270 Chr.1: 109739356-109739356 on Build GRCh38
rs1011817381 Chr.1: 109739446-109739446 on Build GRCh38
rs761100109 Chr.1: 109739538-109739538 on Build GRCh38
rs975121592 Chr.1: 109739569-109739569 on Build GRCh38
rs534810155 Chr.1: 109739565-109739565 on Build GRCh38

View Full Product Details