Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110223544-110223771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC4
Entrez Gene ID
3749
Gene Name
potassium voltage-gated channel subfamily C member 4
Gene Aliases
C1orf30, HKSHIIIC, KSHIIIC, KV3.4
UniGene
Hs.153521
Gene Chromosome Location
Chr.1: 110210714-110282649 on Build GRCh38
Transcripts
NM_001039574.2 NM_004978.4 NR_036437.1 XM_006710625.3 XM_006710627.3 XM_011541401.2 XM_011541402.1 XM_011541403.1 XM_011541404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1583249 c.1305G>A p.T435T Chr.1: 110223590-110223590 on Build GRCh38
4525146 c.1311G>T p.L437L Chr.1: 110223596-110223596 on Build GRCh38
458738 c.1475G>A p.R492Q Chr.1: 110223760-110223760 on Build GRCh38
5594731 c.1392C>T p.I464I Chr.1: 110223677-110223677 on Build GRCh38
2118494 c.1296C>T p.V432V Chr.1: 110223581-110223581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774705792 Chr.1: 110223722-110223722 on Build GRCh38
rs775939037 Chr.1: 110223710-110223710 on Build GRCh38
rs753798744 Chr.1: 110223631-110223631 on Build GRCh38
rs755978329 Chr.1: 110223623-110223623 on Build GRCh38
rs769351147 Chr.1: 110223715-110223715 on Build GRCh38
rs764282706 Chr.1: 110223581-110223581 on Build GRCh38
rs985277923 Chr.1: 110223732-110223732 on Build GRCh38
rs202030640 Chr.1: 110223614-110223614 on Build GRCh38
rs953966957 Chr.1: 110223714-110223714 on Build GRCh38
rs757822089 Chr.1: 110223677-110223677 on Build GRCh38
rs746598938 Chr.1: 110223696-110223696 on Build GRCh38
rs371967697 Chr.1: 110223620-110223620 on Build GRCh38
rs966007199 Chr.1: 110223678-110223678 on Build GRCh38
rs200055497 Chr.1: 110223706-110223706 on Build GRCh38
rs201939187 Chr.1: 110223665-110223665 on Build GRCh38
rs747764524 Chr.1: 110223667-110223667 on Build GRCh38
rs372345369 Chr.1: 110223590-110223590 on Build GRCh38
rs1029005555 Chr.1: 110223747-110223747 on Build GRCh38
rs770301734 Chr.1: 110223699-110223699 on Build GRCh38
rs1000017547 Chr.1: 110223664-110223664 on Build GRCh38
rs761793237 Chr.1: 110223599-110223599 on Build GRCh38
rs549855010 Chr.1: 110223613-110223613 on Build GRCh38
rs367870522 Chr.1: 110223711-110223711 on Build GRCh38
rs145221541 Chr.1: 110223626-110223626 on Build GRCh38
rs375280415 Chr.1: 110223602-110223602 on Build GRCh38
rs777518449 Chr.1: 110223684-110223684 on Build GRCh38
rs191317326 Chr.1: 110223727-110223727 on Build GRCh38

View Full Product Details