Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7890518-7890791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
UniGene
Hs.25601
Gene Chromosome Location
Chr.17: 7884764-7912757 on Build GRCh38
Transcripts
NM_001005271.2 NM_001005273.2 NM_005852.3 XM_005256427.4 XM_005256428.4 XM_005256429.4 XM_005256431.4 XM_006721423.3 XM_006721424.3 XM_006721428.3 XM_017024061.1 XM_017024062.1 XM_017024063.1 XM_017024064.1 XM_017024065.1 XM_017024066.1 XM_017024067.1 XM_017024068.1 XM_017024069.1 XM_017024070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4070327 c.350G>A p.R117Q Chr.17: 7890707-7890707 on Build GRCh38
985421 c.252C>T p.Y84Y Chr.17: 7890609-7890609 on Build GRCh38
985423 c.307C>T p.R103* Chr.17: 7890664-7890664 on Build GRCh38
6440951 c.238G>A p.E80K Chr.17: 7890595-7890595 on Build GRCh38
6571896 c.342_344delGAC p.T115delT Chr.17: 7890699-7890701 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751546224 Chr.17: 7890708-7890708 on Build GRCh38
rs767034177 Chr.17: 7890656-7890656 on Build GRCh38
rs143327592 Chr.17: 7890680-7890680 on Build GRCh38
rs1047123204 Chr.17: 7890670-7890670 on Build GRCh38
rs368862267 Chr.17: 7890699-7890699 on Build GRCh38
rs749820941 Chr.17: 7890751-7890751 on Build GRCh38
rs771266385 Chr.17: 7890665-7890667 on Build GRCh38
rs753770040 Chr.17: 7890697-7890697 on Build GRCh38
rs762376795 Chr.17: 7890642-7890642 on Build GRCh38
rs372004415 Chr.17: 7890560-7890560 on Build GRCh38
rs1053918168 Chr.17: 7890710-7890710 on Build GRCh38
rs751212280 Chr.17: 7890565-7890565 on Build GRCh38
rs758935499 Chr.17: 7890766-7890766 on Build GRCh38
rs188762501 Chr.17: 7890760-7890760 on Build GRCh38
rs184223900 Chr.17: 7890747-7890747 on Build GRCh38
rs770273476 Chr.17: 7890721-7890721 on Build GRCh38
rs923679399 Chr.17: 7890749-7890749 on Build GRCh38
rs778494683 Chr.17: 7890741-7890741 on Build GRCh38
rs771287937 Chr.17: 7890754-7890754 on Build GRCh38
rs748572297 Chr.17: 7890717-7890717 on Build GRCh38
rs747258702 Chr.17: 7890631-7890631 on Build GRCh38
rs999597296 Chr.17: 7890574-7890574 on Build GRCh38
rs777652324 Chr.17: 7890606-7890606 on Build GRCh38
rs549525057 Chr.17: 7890562-7890562 on Build GRCh38
rs767350598 Chr.17: 7890575-7890575 on Build GRCh38
rs781534324 Chr.17: 7890712-7890712 on Build GRCh38
rs758459375 Chr.17: 7890611-7890611 on Build GRCh38
rs138306277 Chr.17: 7890649-7890649 on Build GRCh38
rs902202379 Chr.17: 7890564-7890564 on Build GRCh38
rs1023066121 Chr.17: 7890599-7890599 on Build GRCh38
rs935007033 Chr.17: 7890756-7890756 on Build GRCh38
rs780206250 Chr.17: 7890628-7890628 on Build GRCh38
rs770465240 Chr.17: 7890648-7890648 on Build GRCh38
rs943849975 Chr.17: 7890696-7890696 on Build GRCh38
rs756760970 Chr.17: 7890658-7890663 on Build GRCh38
rs745992945 Chr.17: 7890610-7890610 on Build GRCh38
rs752525306 Chr.17: 7890659-7890659 on Build GRCh38
rs760436956 Chr.17: 7890665-7890665 on Build GRCh38
rs1011702964 Chr.17: 7890595-7890595 on Build GRCh38
rs755261950 Chr.17: 7890709-7890709 on Build GRCh38
rs756074044 Chr.17: 7890590-7890590 on Build GRCh38
rs116087083 Chr.17: 7890654-7890654 on Build GRCh38
rs867251299 Chr.17: 7890712-7890712 on Build GRCh38
rs753823202 Chr.17: 7890548-7890553 on Build GRCh38
rs754798040 Chr.17: 7890568-7890568 on Build GRCh38
rs752437456 Chr.17: 7890582-7890582 on Build GRCh38
rs376670741 Chr.17: 7890698-7890698 on Build GRCh38
rs569669003 Chr.17: 7890632-7890632 on Build GRCh38
rs765997564 Chr.17: 7890555-7890555 on Build GRCh38
rs537435704 Chr.17: 7890540-7890540 on Build GRCh38
rs938168299 Chr.17: 7890701-7890701 on Build GRCh38
rs749962619 Chr.17: 7890692-7890694 on Build GRCh38
rs765578621 Chr.17: 7890653-7890653 on Build GRCh38
rs777077851 Chr.17: 7890641-7890641 on Build GRCh38

View Full Product Details