Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70325525-70325798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX19B
Entrez Gene ID
11269
Gene Name
DEAD-box helicase 19B
Gene Aliases
DBP5, DDX19, RNAh
UniGene
Hs.221761
Gene Chromosome Location
Chr.16: 70289767-70335213 on Build GRCh38
Transcripts
NM_001014449.2 NM_001014451.2 NM_001257172.1 NM_001257173.1 NM_001257174.1 NM_001257175.1 NM_007242.5 XM_006721127.2 XM_011522832.2 XM_011522833.2 XM_011522834.2 XM_017022890.1 NR_039997.1

Gene Symbol
LOC100506083
Entrez Gene ID
100506083
Gene Name
uncharacterized LOC100506083
Gene Aliases
-
UniGene
Hs.635008
Gene Chromosome Location
Chr.16: 70315640-70346747 on Build GRCh38
Transcripts
NM_001014449.2 NM_001014451.2 NM_001257172.1 NM_001257173.1 NM_001257174.1 NM_001257175.1 NM_007242.5 XM_006721127.2 XM_011522832.2 XM_011522833.2 XM_011522834.2 XM_017022890.1 NR_039997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4062328 c.523G>A p.A175T Chr.16: 70325604-70325604 on Build GRCh38
4062326 c.512C>T p.T171M Chr.16: 70325593-70325593 on Build GRCh38
3782982 c.599G>A p.G200D Chr.16: 70325680-70325680 on Build GRCh38
6344487 c.607+5G>T p.? Chr.16: 70325693-70325693 on Build GRCh38
3691309 c.595C>T p.R199* Chr.16: 70325676-70325676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770340872 Chr.16: 70325721-70325721 on Build GRCh38
rs763522884 Chr.16: 70325735-70325735 on Build GRCh38
rs140554740 Chr.16: 70325616-70325616 on Build GRCh38
rs539650364 Chr.16: 70325708-70325708 on Build GRCh38
rs1026400851 Chr.16: 70325575-70325575 on Build GRCh38
rs563060454 Chr.16: 70325716-70325716 on Build GRCh38
rs763472554 Chr.16: 70325737-70325737 on Build GRCh38
rs749257884 Chr.16: 70325667-70325667 on Build GRCh38
rs756280078 Chr.16: 70325622-70325622 on Build GRCh38
rs779949295 Chr.16: 70325665-70325665 on Build GRCh38
rs746959500 Chr.16: 70325713-70325715 on Build GRCh38
rs1008911119 Chr.16: 70325609-70325609 on Build GRCh38
rs977856357 Chr.16: 70325753-70325753 on Build GRCh38
rs777893383 Chr.16: 70325626-70325626 on Build GRCh38
rs375923338 Chr.16: 70325734-70325734 on Build GRCh38
rs551039117 Chr.16: 70325713-70325713 on Build GRCh38
rs773887787 Chr.16: 70325738-70325738 on Build GRCh38
rs1047701495 Chr.16: 70325729-70325729 on Build GRCh38
rs375444457 Chr.16: 70325671-70325671 on Build GRCh38
rs370203489 Chr.16: 70325702-70325702 on Build GRCh38
rs74024199 Chr.16: 70325703-70325703 on Build GRCh38
rs966526042 Chr.16: 70325744-70325744 on Build GRCh38
rs547675730 Chr.16: 70325678-70325678 on Build GRCh38
rs139391794 Chr.16: 70325761-70325761 on Build GRCh38
rs762357250 Chr.16: 70325597-70325597 on Build GRCh38
rs569456895 Chr.16: 70325754-70325754 on Build GRCh38
rs111815770 Chr.16: 70325669-70325669 on Build GRCh38
rs765811423 Chr.16: 70325588-70325588 on Build GRCh38
rs747153226 Chr.16: 70325688-70325688 on Build GRCh38
rs150416272 Chr.16: 70325654-70325654 on Build GRCh38
rs777160984 Chr.16: 70325555-70325555 on Build GRCh38
rs767204262 Chr.16: 70325628-70325628 on Build GRCh38
rs1018049283 Chr.16: 70325657-70325657 on Build GRCh38
rs751106898 Chr.16: 70325604-70325604 on Build GRCh38
rs948053040 Chr.16: 70325699-70325699 on Build GRCh38
rs763676230 Chr.16: 70325600-70325600 on Build GRCh38
rs760031392 Chr.16: 70325558-70325558 on Build GRCh38
rs776132389 Chr.16: 70325592-70325592 on Build GRCh38

View Full Product Details