Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161048213-161048474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP30
Entrez Gene ID
257106
Gene Name
Rho GTPase activating protein 30
Gene Aliases
-
UniGene
Hs.389374
Gene Chromosome Location
Chr.1: 161046942-161069971 on Build GRCh38
Transcripts
NM_001025598.1 NM_001287600.1 NM_001287602.1 NM_181720.2 XM_005245070.2 XM_005245073.3 XM_011509391.2 XM_017000960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5475542 c.2693T>C p.M898T Chr.1: 161048328-161048328 on Build GRCh38
4435058 c.2754T>C p.G918G Chr.1: 161048267-161048267 on Build GRCh38
4435059 c.2121T>C p.G707G Chr.1: 161048267-161048267 on Build GRCh38
133978 c.2572G>A p.E858K Chr.1: 161048449-161048449 on Build GRCh38
2086452 c.2149C>G p.L717V Chr.1: 161048239-161048239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769860057 Chr.1: 161048254-161048254 on Build GRCh38
rs766899947 Chr.1: 161048310-161048310 on Build GRCh38
rs150721972 Chr.1: 161048299-161048299 on Build GRCh38
rs147161320 Chr.1: 161048335-161048335 on Build GRCh38
rs775294757 Chr.1: 161048369-161048369 on Build GRCh38
rs760658374 Chr.1: 161048371-161048371 on Build GRCh38
rs138976951 Chr.1: 161048240-161048240 on Build GRCh38
rs768337789 Chr.1: 161048374-161048374 on Build GRCh38
rs901115166 Chr.1: 161048236-161048236 on Build GRCh38
rs771938023 Chr.1: 161048277-161048277 on Build GRCh38
rs538155601 Chr.1: 161048418-161048418 on Build GRCh38
rs748205506 Chr.1: 161048253-161048253 on Build GRCh38
rs767853682 Chr.1: 161048402-161048402 on Build GRCh38
rs1022424712 Chr.1: 161048370-161048370 on Build GRCh38
rs779069746 Chr.1: 161048362-161048362 on Build GRCh38
rs777821976 Chr.1: 161048349-161048349 on Build GRCh38
rs747682410 Chr.1: 161048448-161048448 on Build GRCh38
rs765012517 Chr.1: 161048390-161048390 on Build GRCh38
rs542240842 Chr.1: 161048408-161048408 on Build GRCh38
rs200004996 Chr.1: 161048406-161048406 on Build GRCh38
rs763630522 Chr.1: 161048296-161048296 on Build GRCh38
rs11587603 Chr.1: 161048417-161048417 on Build GRCh38
rs905277360 Chr.1: 161048372-161048372 on Build GRCh38
rs774114536 Chr.1: 161048263-161048263 on Build GRCh38
rs757281711 Chr.1: 161048410-161048410 on Build GRCh38
rs751939295 Chr.1: 161048316-161048316 on Build GRCh38
rs771844552 Chr.1: 161048368-161048368 on Build GRCh38
rs985743134 Chr.1: 161048411-161048411 on Build GRCh38
rs143839762 Chr.1: 161048288-161048288 on Build GRCh38
rs200666764 Chr.1: 161048238-161048238 on Build GRCh38
rs373597385 Chr.1: 161048431-161048431 on Build GRCh38
rs759561142 Chr.1: 161048264-161048264 on Build GRCh38
rs755329357 Chr.1: 161048318-161048318 on Build GRCh38
rs922995734 Chr.1: 161048387-161048387 on Build GRCh38
rs750040202 Chr.1: 161048392-161048392 on Build GRCh38
rs747576676 Chr.1: 161048230-161048230 on Build GRCh38
rs749442300 Chr.1: 161048259-161048259 on Build GRCh38
rs368020220 Chr.1: 161048437-161048437 on Build GRCh38
rs143251755 Chr.1: 161048434-161048434 on Build GRCh38
rs779122532 Chr.1: 161048412-161048412 on Build GRCh38
rs892432863 Chr.1: 161048311-161048311 on Build GRCh38
rs761597646 Chr.1: 161048389-161048389 on Build GRCh38
rs775449947 Chr.1: 161048285-161048285 on Build GRCh38
rs771113732 Chr.1: 161048358-161048358 on Build GRCh38
rs781570242 Chr.1: 161048325-161048325 on Build GRCh38
rs776670329 Chr.1: 161048442-161048442 on Build GRCh38
rs745673736 Chr.1: 161048366-161048366 on Build GRCh38
rs137873621 Chr.1: 161048300-161048300 on Build GRCh38
rs532517541 Chr.1: 161048400-161048400 on Build GRCh38
rs967200766 Chr.1: 161048338-161048338 on Build GRCh38
rs776256738 Chr.1: 161048379-161048379 on Build GRCh38
rs868579432 Chr.1: 161048323-161048323 on Build GRCh38
rs201040129 Chr.1: 161048424-161048424 on Build GRCh38
rs749565030 Chr.1: 161048354-161048354 on Build GRCh38

View Full Product Details