Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 160129021-160129260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A2
Entrez Gene ID
477
Gene Name
ATPase Na+/K+ transporting subunit alpha 2
Gene Aliases
FHM2, MHP2
UniGene
Hs.34114
Gene Chromosome Location
Chr.1: 160115730-160143591 on Build GRCh38
Transcripts
NM_000702.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
898169 c.1262G>A p.R421Q Chr.1: 160129025-160129025 on Build GRCh38
5916987 c.1316C>T p.S439F Chr.1: 160129079-160129079 on Build GRCh38
1335455 c.1282C>T p.R428C Chr.1: 160129045-160129045 on Build GRCh38
2085186 c.1287C>T p.A429A Chr.1: 160129050-160129050 on Build GRCh38
3476747 c.1317C>T p.S439S Chr.1: 160129080-160129080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397978917 Chr.1: 160129140-160129140 on Build GRCh38
rs56220830 Chr.1: 160129140-160129141 on Build GRCh38
rs983157919 Chr.1: 160129124-160129124 on Build GRCh38
rs1057322889 Chr.1: 160129198-160129198 on Build GRCh38
rs373294271 Chr.1: 160129108-160129108 on Build GRCh38
rs370111257 Chr.1: 160129051-160129051 on Build GRCh38
rs201515950 Chr.1: 160129220-160129220 on Build GRCh38
rs746478136 Chr.1: 160129046-160129046 on Build GRCh38
rs1043259611 Chr.1: 160129228-160129228 on Build GRCh38
rs922089591 Chr.1: 160129129-160129129 on Build GRCh38
rs946293726 Chr.1: 160129199-160129199 on Build GRCh38
rs780843759 Chr.1: 160129098-160129098 on Build GRCh38
rs375905731 Chr.1: 160129126-160129126 on Build GRCh38
rs1003846681 Chr.1: 160129076-160129076 on Build GRCh38
rs759939624 Chr.1: 160129080-160129080 on Build GRCh38
rs373024075 Chr.1: 160129226-160129226 on Build GRCh38
rs953524389 Chr.1: 160129149-160129149 on Build GRCh38
rs780819805 Chr.1: 160129215-160129215 on Build GRCh38
rs752479774 Chr.1: 160129047-160129047 on Build GRCh38
rs397801520 Chr.1: 160129138-160129139 on Build GRCh38
rs763770380 Chr.1: 160129071-160129071 on Build GRCh38
rs746959685 Chr.1: 160129139-160129139 on Build GRCh38
rs530469412 Chr.1: 160129140-160129140 on Build GRCh38
rs778544889 Chr.1: 160129113-160129113 on Build GRCh38
rs542189014 Chr.1: 160129102-160129103 on Build GRCh38
rs763762298 Chr.1: 160129093-160129093 on Build GRCh38
rs776003260 Chr.1: 160129220-160129220 on Build GRCh38
rs866493442 Chr.1: 160129069-160129069 on Build GRCh38
rs772571357 Chr.1: 160129230-160129230 on Build GRCh38
rs17846711 Chr.1: 160129139-160129139 on Build GRCh38
rs772762868 Chr.1: 160129222-160129222 on Build GRCh38
rs753995041 Chr.1: 160129102-160129102 on Build GRCh38
rs764284991 Chr.1: 160129100-160129100 on Build GRCh38
rs776086125 Chr.1: 160129048-160129048 on Build GRCh38
rs544710713 Chr.1: 160129114-160129114 on Build GRCh38
rs762571297 Chr.1: 160129050-160129050 on Build GRCh38
rs912112552 Chr.1: 160129176-160129176 on Build GRCh38
rs751078196 Chr.1: 160129096-160129096 on Build GRCh38
rs201570970 Chr.1: 160129137-160129137 on Build GRCh38
rs768088341 Chr.1: 160129081-160129081 on Build GRCh38
rs938896598 Chr.1: 160129170-160129170 on Build GRCh38
rs374067427 Chr.1: 160129060-160129060 on Build GRCh38
rs776084261 Chr.1: 160129234-160129234 on Build GRCh38
rs545633334 Chr.1: 160129156-160129156 on Build GRCh38
rs5778151 Chr.1: 160129137-160129138 on Build GRCh38
rs975166744 Chr.1: 160129119-160129119 on Build GRCh38
rs779985796 Chr.1: 160129045-160129045 on Build GRCh38
rs777558964 Chr.1: 160129070-160129072 on Build GRCh38
rs769533979 Chr.1: 160129221-160129221 on Build GRCh38

View Full Product Details