Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 110498259-110498508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP5
Entrez Gene ID
1847
Gene Name
dual specificity phosphatase 5
Gene Aliases
DUSP, HVH3
UniGene
Hs.2128
Gene Chromosome Location
Chr.10: 110497867-110511544 on Build GRCh38
Transcripts
NM_004419.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6205420 c.229C>G p.Q77E Chr.10: 110498350-110498350 on Build GRCh38
1602964 c.356G>T p.G119V Chr.10: 110498477-110498477 on Build GRCh38
6571213 c.141C>G p.L47L Chr.10: 110498262-110498262 on Build GRCh38
5049887 c.168G>T p.R56R Chr.10: 110498289-110498289 on Build GRCh38
6128561 c.183G>T p.S61S Chr.10: 110498304-110498304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752833763 Chr.10: 110498460-110498460 on Build GRCh38
rs779141439 Chr.10: 110498282-110498282 on Build GRCh38
rs779885359 Chr.10: 110498382-110498382 on Build GRCh38
rs771904395 Chr.10: 110498431-110498431 on Build GRCh38
rs12413389 Chr.10: 110498374-110498374 on Build GRCh38
rs996195449 Chr.10: 110498373-110498373 on Build GRCh38
rs753917795 Chr.10: 110498322-110498322 on Build GRCh38
rs992424271 Chr.10: 110498479-110498479 on Build GRCh38
rs775042496 Chr.10: 110498291-110498291 on Build GRCh38
rs765388886 Chr.10: 110498334-110498334 on Build GRCh38
rs758687361 Chr.10: 110498354-110498354 on Build GRCh38
rs560255020 Chr.10: 110498414-110498414 on Build GRCh38
rs1031351877 Chr.10: 110498450-110498450 on Build GRCh38
rs1052794501 Chr.10: 110498347-110498347 on Build GRCh38
rs925579468 Chr.10: 110498344-110498344 on Build GRCh38
rs762408672 Chr.10: 110498295-110498295 on Build GRCh38
rs756046182 Chr.10: 110498377-110498377 on Build GRCh38
rs773599328 Chr.10: 110498435-110498435 on Build GRCh38
rs748756210 Chr.10: 110498388-110498388 on Build GRCh38
rs527596389 Chr.10: 110498442-110498442 on Build GRCh38
rs542883088 Chr.10: 110498445-110498445 on Build GRCh38
rs761226631 Chr.10: 110498313-110498313 on Build GRCh38
rs774077983 Chr.10: 110498394-110498394 on Build GRCh38
rs773512860 Chr.10: 110498312-110498312 on Build GRCh38
rs780173888 Chr.10: 110498278-110498278 on Build GRCh38
rs185835786 Chr.10: 110498457-110498457 on Build GRCh38
rs979866130 Chr.10: 110498292-110498292 on Build GRCh38
rs765298963 Chr.10: 110498458-110498458 on Build GRCh38
rs755076416 Chr.10: 110498330-110498330 on Build GRCh38
rs376373405 Chr.10: 110498475-110498475 on Build GRCh38
rs750191676 Chr.10: 110498477-110498477 on Build GRCh38
rs375225612 Chr.10: 110498286-110498286 on Build GRCh38
rs768453959 Chr.10: 110498355-110498356 on Build GRCh38
rs774167871 Chr.10: 110498296-110498296 on Build GRCh38
rs971471118 Chr.10: 110498406-110498406 on Build GRCh38
rs752921905 Chr.10: 110498341-110498341 on Build GRCh38
rs577810725 Chr.10: 110498355-110498355 on Build GRCh38
rs753918329 Chr.10: 110498281-110498281 on Build GRCh38
rs746760178 Chr.10: 110498283-110498283 on Build GRCh38
rs755882591 Chr.10: 110498480-110498480 on Build GRCh38
rs545263710 Chr.10: 110498361-110498361 on Build GRCh38
rs768292551 Chr.10: 110498393-110498393 on Build GRCh38
rs548929241 Chr.10: 110498451-110498451 on Build GRCh38
rs935640143 Chr.10: 110498346-110498346 on Build GRCh38
rs748698192 Chr.10: 110498332-110498333 on Build GRCh38
rs760979839 Chr.10: 110498441-110498441 on Build GRCh38
rs758597468 Chr.10: 110498467-110498467 on Build GRCh38
rs759547782 Chr.10: 110498454-110498454 on Build GRCh38
rs200429102 Chr.10: 110498288-110498288 on Build GRCh38
rs762208561 Chr.10: 110498308-110498308 on Build GRCh38
rs369887116 Chr.10: 110498287-110498287 on Build GRCh38
rs74386806 Chr.10: 110498284-110498284 on Build GRCh38
rs756078547 Chr.10: 110498277-110498277 on Build GRCh38

View Full Product Details